Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
ENST00000331163.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
ENST00000331163.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
ENST00000331163.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
ENST00000331163.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
ENST00000331163.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
ENST00000331163.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
ENST00000331163.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
ENST00000331163.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
ENST00000331163.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
ENST00000331163.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
ENST00000331163.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
ENST00000331163.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
ENST00000331163.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
ENST00000331163.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
ENST00000331163.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
ENST00000381551.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
ENST00000381551.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
ENST00000381551.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
ENST00000381551.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
ENST00000381551.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
ENST00000381551.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
ENST00000381551.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
ENST00000381551.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
ENST00000381551.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
ENST00000381551.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
ENST00000381551.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
ENST00000440375.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
ENST00000440375.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
ENST00000440375.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
ENST00000440375.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
ENST00000440375.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
ENST00000455790.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
ENST00000455790.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
ENST00000455790.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
ENST00000455790.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
ENST00000455790.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
ENST00000455790.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
NM_002608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
NM_002608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
NM_002608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
NM_002608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
NM_002608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
NM_002608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
NM_002608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
NM_002608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
NM_002608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
NM_002608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
NM_002608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
NM_002608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
NM_002608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
NM_002608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
NM_002608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
NM_002608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
NM_033016.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
NM_033016.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
NM_033016.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
NM_033016.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
NM_033016.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
NM_033016.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
NM_033016.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
NM_033016.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
NM_033016.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
NM_033016.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
NM_033016.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008741.hg.1 |
NM_033016.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22013617.hg.1 |
ENST00000331163.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22013617.hg.1 |
ENST00000381551.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22013617.hg.1 |
NM_002608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22013617.hg.1 |
NM_033016.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22013617.hg.1 |
TC22000802.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2200163389.hg.1 |
TC2200008741.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200163389.hg.1 |
ENST00000331163.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200163389.hg.1 |
ENST00000381551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200163389.hg.1 |
ENST00000440375.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200163389.hg.1 |
ENST00000455790.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200163389.hg.1 |
NM_002608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200163389.hg.1 |
NM_033016.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200163389.hg.1 |
ENST00000331163.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200163389.hg.1 |
ENST00000381551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200163389.hg.1 |
ENST00000440375.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200163389.hg.1 |
ENST00000455790.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200163389.hg.1 |
NM_002608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200163389.hg.1 |
NM_033016.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
ENST00000331163.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
ENST00000331163.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
ENST00000331163.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
ENST00000331163.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
ENST00000331163.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
ENST00000331163.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
ENST00000331163.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
ENST00000331163.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
ENST00000331163.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
ENST00000331163.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
ENST00000331163.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
ENST00000331163.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
ENST00000331163.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
ENST00000331163.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
ENST00000331163.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
ENST00000381551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
ENST00000381551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
ENST00000381551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
ENST00000381551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
ENST00000381551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
ENST00000381551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
ENST00000381551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
ENST00000381551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
ENST00000381551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
ENST00000381551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
ENST00000381551.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
ENST00000440375.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
ENST00000440375.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
ENST00000440375.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
ENST00000440375.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
ENST00000440375.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
ENST00000455790.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
ENST00000455790.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
ENST00000455790.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
ENST00000455790.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
ENST00000455790.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
ENST00000455790.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
NM_002608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
NM_002608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
NM_002608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
NM_002608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
NM_002608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
NM_002608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
NM_002608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
NM_002608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
NM_002608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
NM_002608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
NM_002608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
NM_002608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
NM_002608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
NM_002608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
NM_002608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
NM_002608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
NM_033016.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
NM_033016.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
NM_033016.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
NM_033016.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
NM_033016.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
NM_033016.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
NM_033016.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
NM_033016.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
NM_033016.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
NM_033016.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
NM_033016.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008741.hg.1 |
NM_033016.3 |
Refseq |