Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009755.hg.1 |
ENST00000252997.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009755.hg.1 |
ENST00000252997.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009755.hg.1 |
ENST00000252997.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009755.hg.1 |
ENST00000252997.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009755.hg.1 |
ENST00000252997.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009755.hg.1 |
ENST00000252997.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009755.hg.1 |
ENST00000252997.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009755.hg.1 |
ENST00000252997.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009755.hg.1 |
ENST00000252997.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009755.hg.1 |
NM_080473.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009755.hg.1 |
NM_080473.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009755.hg.1 |
NM_080473.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009755.hg.1 |
NM_080473.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009755.hg.1 |
NM_080473.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009755.hg.1 |
NM_080473.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009755.hg.1 |
NM_080473.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009755.hg.1 |
NM_080473.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009755.hg.1 |
NM_080473.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013886.hg.1 |
ENST00000252997.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013886.hg.1 |
ENST00000370545.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013886.hg.1 |
ENST00000540404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013886.hg.1 |
NM_080473.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013886.hg.1 |
TC20001011.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000168048.hg.1 |
ENST00000252997.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168048.hg.1 |
NM_080473.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168048.hg.1 |
TC2000009755.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000168048.hg.1 |
ENST00000252997.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168048.hg.1 |
NM_080473.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009755.hg.1 |
ENST00000252997.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009755.hg.1 |
ENST00000252997.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009755.hg.1 |
ENST00000252997.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009755.hg.1 |
ENST00000252997.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009755.hg.1 |
ENST00000252997.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009755.hg.1 |
ENST00000252997.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009755.hg.1 |
ENST00000252997.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009755.hg.1 |
ENST00000252997.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009755.hg.1 |
ENST00000252997.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009755.hg.1 |
NM_080473.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009755.hg.1 |
NM_080473.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009755.hg.1 |
NM_080473.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009755.hg.1 |
NM_080473.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009755.hg.1 |
NM_080473.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009755.hg.1 |
NM_080473.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009755.hg.1 |
NM_080473.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009755.hg.1 |
NM_080473.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009755.hg.1 |
NM_080473.4 |
Refseq |