Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022130.hg.1 |
TC09001729.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022130.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022130.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900182013.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182013.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900182013.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900182013.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900182013.hg.1 |
TC0900011976.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
ENST00000298532.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011976.hg.1 |
NM_003086.3 |
Refseq |