Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | X81882.1 | - | 19 - 20 | 2948 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Homo sapiens Probe spans exons |