Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000354431.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000354431.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000354431.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000354431.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000354431.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000354431.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000354431.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000354431.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000354431.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000354431.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000354431.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000354431.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000354431.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000354431.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000370909.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000370909.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000370909.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000370909.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000370909.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000370909.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000370911.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000370911.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000370911.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000370911.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000370911.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000370911.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000370911.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000395089.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000395089.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000395089.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000395089.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000395089.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000395089.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000395089.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000395089.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000395089.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000395089.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000395089.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000395089.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000415760.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000415760.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000415760.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000415760.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000415760.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000415760.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000415760.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000415760.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000415760.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000415760.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000415760.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000415760.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000415760.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000415760.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000415760.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000415760.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000417067.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000417067.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000417067.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000417067.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000444984.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000444984.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000444984.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000463724.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000463724.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000463724.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000463724.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000463724.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000463724.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000463724.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000463724.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000468759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000468759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000468759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000468759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000468759.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000495179.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000495179.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
ENST00000495179.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
NM_001105659.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
NM_001105659.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
NM_001105659.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
NM_001105659.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
NM_001105659.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
NM_001105659.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
NM_001105659.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
NM_001105659.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
NM_001105659.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
NM_001105659.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
NM_001105659.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
NM_001105659.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
NM_001105659.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014605.hg.1 |
NM_001105659.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000354431.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000354431.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000354431.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000354431.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000354431.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000354431.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000354431.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000354431.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000354431.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000354431.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000354431.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000354431.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000354431.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000354431.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000370909.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000370909.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000370909.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000370909.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000370909.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000370909.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000370911.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000370911.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000370911.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000370911.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000370911.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000370911.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000370911.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000395089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000395089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000395089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000395089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000395089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000395089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000395089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000395089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000395089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000395089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000395089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000395089.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000415760.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000415760.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000415760.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000415760.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000415760.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000415760.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000415760.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000415760.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000415760.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000415760.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000415760.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000415760.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000415760.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000415760.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000415760.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000415760.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000417067.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000417067.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000417067.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000417067.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000444984.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000444984.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000444984.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000463724.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000463724.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000463724.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000463724.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000463724.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000463724.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000463724.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000463724.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000468759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000468759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000468759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000468759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000468759.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000495179.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000495179.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
ENST00000495179.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
NM_001105659.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
NM_001105659.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
NM_001105659.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
NM_001105659.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
NM_001105659.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
NM_001105659.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
NM_001105659.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
NM_001105659.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
NM_001105659.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
NM_001105659.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
NM_001105659.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
NM_001105659.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
NM_001105659.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100014605.hg.1 |
NM_001105659.1 |
Refseq |