Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054270.hg.1 |
TC01003491.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054270.hg.1 |
ENST00000367817.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054270.hg.1 |
NM_001937.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016339.hg.1 |
ENST00000367817.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016339.hg.1 |
ENST00000367817.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016339.hg.1 |
ENST00000367817.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016339.hg.1 |
ENST00000367817.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016339.hg.1 |
ENST00000367817.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016339.hg.1 |
ENST00000367817.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016339.hg.1 |
NM_001937.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016339.hg.1 |
NM_001937.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016339.hg.1 |
NM_001937.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016339.hg.1 |
NM_001937.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016339.hg.1 |
NM_001937.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054271.hg.1 |
TC01003491.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054271.hg.1 |
ENST00000367817.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054271.hg.1 |
NM_001937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100224248.hg.1 |
ENST00000367817.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100224248.hg.1 |
NM_001937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100224248.hg.1 |
ENST00000367817.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100224248.hg.1 |
NM_001937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100224248.hg.1 |
TC0100016339.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100224249.hg.1 |
ENST00000367817.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100224249.hg.1 |
NM_001937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100224249.hg.1 |
ENST00000367817.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100224249.hg.1 |
NM_001937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100224249.hg.1 |
TC0100016339.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100016339.hg.1 |
ENST00000367817.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016339.hg.1 |
ENST00000367817.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016339.hg.1 |
ENST00000367817.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016339.hg.1 |
ENST00000367817.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016339.hg.1 |
ENST00000367817.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016339.hg.1 |
ENST00000367817.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016339.hg.1 |
NM_001937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016339.hg.1 |
NM_001937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016339.hg.1 |
NM_001937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016339.hg.1 |
NM_001937.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016339.hg.1 |
NM_001937.4 |
Refseq |