Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026681.hg.1 |
ENST00000279441.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026681.hg.1 |
NM_002425.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026681.hg.1 |
TC11002233.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026683.hg.1 |
ENST00000279441.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026683.hg.1 |
NM_002425.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026683.hg.1 |
TC11002233.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026682.hg.1 |
ENST00000279441.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026682.hg.1 |
NM_002425.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11026682.hg.1 |
TC11002233.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012130.hg.1 |
ENST00000279441.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012130.hg.1 |
ENST00000279441.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012130.hg.1 |
ENST00000279441.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012130.hg.1 |
ENST00000279441.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012130.hg.1 |
ENST00000279441.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012130.hg.1 |
ENST00000279441.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012130.hg.1 |
ENST00000279441.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012130.hg.1 |
ENST00000279441.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012130.hg.1 |
ENST00000279441.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012130.hg.1 |
ENST00000279441.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012130.hg.1 |
ENST00000279441.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012130.hg.1 |
ENST00000279441.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012130.hg.1 |
ENST00000279441.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012130.hg.1 |
ENST00000539681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012130.hg.1 |
ENST00000539681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012130.hg.1 |
ENST00000539681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012130.hg.1 |
ENST00000539681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012130.hg.1 |
NM_002425.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012130.hg.1 |
NM_002425.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012130.hg.1 |
NM_002425.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012130.hg.1 |
NM_002425.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012130.hg.1 |
NM_002425.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012130.hg.1 |
NM_002425.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012130.hg.1 |
NM_002425.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012130.hg.1 |
NM_002425.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012130.hg.1 |
NM_002425.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012130.hg.1 |
NM_002425.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012130.hg.1 |
NM_002425.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012130.hg.1 |
NM_002425.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012130.hg.1 |
NM_002425.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193236.hg.1 |
ENST00000279441.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193236.hg.1 |
ENST00000539681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193236.hg.1 |
NM_002425.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193236.hg.1 |
ENST00000279441.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193236.hg.1 |
ENST00000539681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193236.hg.1 |
NM_002425.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193236.hg.1 |
TC1100012130.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100012130.hg.1 |
ENST00000279441.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012130.hg.1 |
ENST00000279441.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012130.hg.1 |
ENST00000279441.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012130.hg.1 |
ENST00000279441.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012130.hg.1 |
ENST00000279441.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012130.hg.1 |
ENST00000279441.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012130.hg.1 |
ENST00000279441.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012130.hg.1 |
ENST00000279441.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012130.hg.1 |
ENST00000279441.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012130.hg.1 |
ENST00000279441.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012130.hg.1 |
ENST00000279441.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012130.hg.1 |
ENST00000279441.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012130.hg.1 |
ENST00000279441.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012130.hg.1 |
ENST00000539681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012130.hg.1 |
ENST00000539681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012130.hg.1 |
ENST00000539681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012130.hg.1 |
ENST00000539681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012130.hg.1 |
NM_002425.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012130.hg.1 |
NM_002425.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012130.hg.1 |
NM_002425.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012130.hg.1 |
NM_002425.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012130.hg.1 |
NM_002425.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012130.hg.1 |
NM_002425.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012130.hg.1 |
NM_002425.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012130.hg.1 |
NM_002425.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012130.hg.1 |
NM_002425.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012130.hg.1 |
NM_002425.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012130.hg.1 |
NM_002425.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012130.hg.1 |
NM_002425.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012130.hg.1 |
NM_002425.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193237.hg.1 |
ENST00000279441.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193237.hg.1 |
ENST00000539681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193237.hg.1 |
NM_002425.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193237.hg.1 |
ENST00000279441.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193237.hg.1 |
ENST00000539681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193237.hg.1 |
NM_002425.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193237.hg.1 |
TC1100012130.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100193238.hg.1 |
ENST00000279441.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193238.hg.1 |
ENST00000539681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193238.hg.1 |
NM_002425.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193238.hg.1 |
ENST00000279441.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193238.hg.1 |
ENST00000539681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100193238.hg.1 |
NM_002425.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100193238.hg.1 |
TC1100012130.hg.1 |
Clariom D TC |