Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03028406.hg.1 |
TC03001562.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03028406.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03028406.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03028407.hg.1 |
TC03001562.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03028407.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03028407.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
ENST00000477856.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
ENST00000477856.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
ENST00000477856.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
ENST00000477856.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300186475.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300186475.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300186475.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300186475.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300186475.hg.1 |
TC0300011566.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
ENST00000477856.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
ENST00000477856.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
ENST00000477856.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
ENST00000477856.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011566.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300186476.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300186476.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300186476.hg.1 |
ENST00000263665.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300186476.hg.1 |
NM_020872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300186476.hg.1 |
TC0300011566.hg.1 |
Clariom D TC |