Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02015611.hg.1 |
TC02001049.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02015611.hg.1 |
ENST00000392539.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02015611.hg.1 |
NM_000523.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02015612.hg.1 |
TC02001049.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02015612.hg.1 |
ENST00000392539.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02015612.hg.1 |
NM_000523.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010045.hg.1 |
ENST00000392539.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010045.hg.1 |
ENST00000392539.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010045.hg.1 |
ENST00000392539.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010045.hg.1 |
NM_000523.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010045.hg.1 |
NM_000523.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010045.hg.1 |
NM_000523.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010045.hg.1 |
NM_000523.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02015609.hg.1 |
TC02001049.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02015609.hg.1 |
NM_000523.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200170468.hg.1 |
ENST00000392539.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200170468.hg.1 |
NM_000523.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200170468.hg.1 |
ENST00000392539.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200170468.hg.1 |
NM_000523.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200170468.hg.1 |
TC0200010045.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200010045.hg.1 |
ENST00000392539.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010045.hg.1 |
ENST00000392539.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010045.hg.1 |
ENST00000392539.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010045.hg.1 |
NM_000523.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010045.hg.1 |
NM_000523.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010045.hg.1 |
NM_000523.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010045.hg.1 |
NM_000523.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200170467.hg.1 |
ENST00000392539.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200170467.hg.1 |
NM_000523.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200170467.hg.1 |
ENST00000392539.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200170467.hg.1 |
NM_000523.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200170467.hg.1 |
TC0200010045.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200170465.hg.1 |
NM_000523.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200170465.hg.1 |
NM_000523.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200170465.hg.1 |
TC0200010045.hg.1 |
Clariom D TC |