Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271087.1 | NP_001258016.1 | 1 - 2 | 209 | 78 | |
NM_001271088.1 | NP_001258017.1 | 1 - 2 | 209 | 78 | ||
NM_005997.2 | NP_005988.1 | 1 - 2 | 209 | 78 | ||
GenBank mRNA | AK225130.1 | - | 1 - 2 | 167 | 78 | |
AK297565.1 | - | 1 - 2 | 171 | 78 | ||
AK310045.1 | - | 1 - 2 | 183 | 78 | ||
BC003151.1 | - | 1 - 2 | 151 | 2958630 | 78 | |
BT019356.1 | - | 1 - 2 | 117 | 78 | ||
BT019357.1 | - | 1 - 2 | 117 | 78 | ||
CT003766.1 | - | 1 - 2 | 141 | 78 | ||
CX163016.1 | - | 1 - 2 | 154 | 78 | ||
D43642.1 | - | 1 - 2 | 160 | 78 | ||
DQ655941.2 | - | 1 - 2 | 210 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |