Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000265000.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000265000.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000265000.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000265000.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000265000.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000265000.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000265000.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000265000.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000265000.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000265000.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000265000.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000265000.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000265000.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000265000.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000265000.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000265000.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000265000.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000265000.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000502407.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000502407.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000502407.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000502407.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000503213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000503213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000503213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000503213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000503213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000503213.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000505308.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000505308.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000505308.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000505308.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000505308.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000505308.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000505308.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000505308.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000505308.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000505308.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000505308.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000505308.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000506317.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000506317.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000506317.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000506317.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000506317.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000512285.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000512285.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000512285.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000512285.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000512285.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000512285.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000512285.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000512285.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000512285.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000512285.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000512285.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000515862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000515862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000515862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000515862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000515862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
ENST00000515862.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
NM_017423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
NM_017423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
NM_017423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
NM_017423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
NM_017423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
NM_017423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
NM_017423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
NM_017423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
NM_017423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
NM_017423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
NM_017423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
NM_017423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
NM_017423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
NM_017423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
NM_017423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
NM_017423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
NM_017423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009322.hg.1 |
NM_017423.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011714.hg.1 |
TC04000853.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011714.hg.1 |
ENST00000265000.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011714.hg.1 |
ENST00000458613.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011714.hg.1 |
ENST00000502407.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011714.hg.1 |
ENST00000512285.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04011714.hg.1 |
NM_017423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163945.hg.1 |
ENST00000265000.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163945.hg.1 |
ENST00000502407.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163945.hg.1 |
ENST00000505308.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163945.hg.1 |
ENST00000512285.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163945.hg.1 |
NM_017423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163945.hg.1 |
ENST00000265000.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163945.hg.1 |
ENST00000502407.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163945.hg.1 |
ENST00000505308.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163945.hg.1 |
ENST00000512285.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400163945.hg.1 |
NM_017423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400163945.hg.1 |
TC0400009322.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000265000.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000265000.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000265000.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000265000.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000265000.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000265000.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000265000.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000265000.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000265000.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000265000.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000265000.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000265000.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000265000.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000265000.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000265000.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000265000.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000265000.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000265000.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000502407.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000502407.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000502407.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000502407.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000503213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000503213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000503213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000503213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000503213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000503213.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000505308.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000505308.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000505308.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000505308.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000505308.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000505308.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000505308.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000505308.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000505308.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000505308.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000505308.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000505308.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000506317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000506317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000506317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000506317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000506317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000512285.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000512285.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000512285.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000512285.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000512285.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000512285.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000512285.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000512285.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000512285.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000512285.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000512285.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000515862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000515862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000515862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000515862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000515862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
ENST00000515862.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
NM_017423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
NM_017423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
NM_017423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
NM_017423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
NM_017423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
NM_017423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
NM_017423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
NM_017423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
NM_017423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
NM_017423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
NM_017423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
NM_017423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
NM_017423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
NM_017423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
NM_017423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
NM_017423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
NM_017423.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009322.hg.1 |
NM_017423.2 |
Refseq |