Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
ENST00000532066.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
ENST00000532066.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
ENST00000532066.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
ENST00000532066.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
ENST00000570295.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151429.hg.1 |
ENST00000299413.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151429.hg.1 |
ENST00000532066.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151429.hg.1 |
NM_017583.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151429.hg.1 |
ENST00000299413.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151429.hg.1 |
ENST00000532066.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100151429.hg.1 |
NM_017583.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100151429.hg.1 |
TC1100007287.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
ENST00000299413.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
ENST00000532066.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
ENST00000532066.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
ENST00000532066.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
ENST00000532066.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
ENST00000570295.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100007287.hg.1 |
NM_017583.5 |
Refseq |