Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000380321.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000380321.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000380321.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000380321.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000380321.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000380321.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000380321.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000380321.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000380321.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000380323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000380323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000380323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000380323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000380323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000380323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000380323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000380323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000380323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000380323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000380323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000380323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000380338.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000380338.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000380338.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000380338.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000380338.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000380338.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000380338.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000380338.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000380338.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000380338.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000380338.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000380338.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000380338.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000380338.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000380338.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000380338.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000380338.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000380338.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000380338.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000380338.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000468513.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000468513.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000468513.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000468513.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000468513.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000468513.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000469261.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000469261.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000469261.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000469261.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000469261.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000469261.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000475957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000475957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000475957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000475957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000475957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000475957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000475957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000488705.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000488705.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000488705.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000488705.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000488705.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000488705.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000491137.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000491137.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000491137.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000491137.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000491137.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000491137.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000629733.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000629733.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000629733.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000629733.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000629733.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000629733.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000629733.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000629733.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000629733.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000629733.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000629733.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000629733.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000629733.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000629733.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000629733.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000629733.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000629733.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000630269.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000630269.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000630269.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000630269.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000630269.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000630269.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000630269.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000630269.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000630269.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000630269.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000630269.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000630269.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000630269.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000630269.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000630269.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
ENST00000630269.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
NM_001286691.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
NM_001286691.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
NM_001286691.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
NM_001286691.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
NM_001286691.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
NM_001286691.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
NM_001286691.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
NM_001286691.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
NM_001286691.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
NM_001286691.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
NM_001286691.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
NM_001286691.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
NM_001286691.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
NM_001286691.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
NM_001286691.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
NM_001286691.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
NM_001286691.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
NM_001286691.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
NM_001286691.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
NM_004529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
NM_004529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
NM_004529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
NM_004529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
NM_004529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
NM_004529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
NM_004529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
NM_004529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
NM_004529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
NM_004529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
NM_004529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
NM_004529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
NM_004529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
NM_004529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
NM_004529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
NM_004529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
NM_004529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
NM_004529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
NM_004529.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900009673.hg.1 |
NM_004529.3 |
Refseq |