Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08010884.hg.1 |
TC08000820.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08010884.hg.1 |
ENST00000289013.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08010884.hg.1 |
NM_052924.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000289013.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000289013.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000289013.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000289013.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000289013.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000289013.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000289013.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000289013.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000289013.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000289013.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000289013.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000289013.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000289013.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000289013.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000289013.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000289013.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000289013.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000289013.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000522335.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000522335.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000522335.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000522335.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000522335.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000522335.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000522335.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000522335.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000522335.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000522335.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000522335.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000522335.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000522335.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000522335.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000522335.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000522335.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000522335.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000522335.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000522899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000522899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000522899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000522899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000522899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000522899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000522899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000522899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000522899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000522899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000620174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000620174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000620174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000620174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000620174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000620174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000620174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000620174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000620174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000620174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000620174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000620174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000620174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000620174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000620174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000620174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000620174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000620174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000620174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
ENST00000620174.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
NM_052924.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
NM_052924.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
NM_052924.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
NM_052924.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
NM_052924.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
NM_052924.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
NM_052924.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
NM_052924.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
NM_052924.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
NM_052924.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
NM_052924.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
NM_052924.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
NM_052924.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
NM_052924.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
NM_052924.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
NM_052924.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
NM_052924.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
NM_052924.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800009194.hg.1 |
NM_052924.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800162577.hg.1 |
ENST00000289013.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800162577.hg.1 |
ENST00000522335.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800162577.hg.1 |
ENST00000620174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800162577.hg.1 |
NM_052924.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800162577.hg.1 |
ENST00000289013.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800162577.hg.1 |
ENST00000522335.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800162577.hg.1 |
ENST00000620174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800162577.hg.1 |
NM_052924.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800162577.hg.1 |
TC0800009194.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000289013.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000289013.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000289013.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000289013.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000289013.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000289013.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000289013.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000289013.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000289013.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000289013.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000289013.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000289013.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000289013.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000289013.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000289013.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000289013.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000289013.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000289013.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000522335.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000522335.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000522335.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000522335.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000522335.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000522335.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000522335.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000522335.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000522335.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000522335.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000522335.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000522335.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000522335.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000522335.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000522335.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000522335.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000522335.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000522335.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000522899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000522899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000522899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000522899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000522899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000522899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000522899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000522899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000522899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000522899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000620174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000620174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000620174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000620174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000620174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000620174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000620174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000620174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000620174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000620174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000620174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000620174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000620174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000620174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000620174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000620174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000620174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000620174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000620174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
ENST00000620174.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
NM_052924.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
NM_052924.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
NM_052924.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
NM_052924.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
NM_052924.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
NM_052924.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
NM_052924.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
NM_052924.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
NM_052924.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
NM_052924.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
NM_052924.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
NM_052924.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
NM_052924.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
NM_052924.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
NM_052924.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
NM_052924.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
NM_052924.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
NM_052924.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800009194.hg.1 |
NM_052924.2 |
Refseq |