Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
ENST00000246020.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
ENST00000246020.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
ENST00000246020.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
ENST00000246020.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
ENST00000246020.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
ENST00000246020.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
ENST00000347397.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
ENST00000347397.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
ENST00000347397.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
ENST00000347397.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
ENST00000347397.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
ENST00000347397.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
ENST00000347397.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
ENST00000347397.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
ENST00000347397.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
ENST00000472140.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
ENST00000472140.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
ENST00000472140.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
ENST00000472140.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
ENST00000472140.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
ENST00000472140.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
ENST00000472140.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
ENST00000485969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
ENST00000485969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
ENST00000485969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
ENST00000485969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
ENST00000485969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
ENST00000485969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
ENST00000619238.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
ENST00000619238.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
ENST00000619238.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
ENST00000619238.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
ENST00000619238.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
ENST00000619238.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
NM_138283.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
NM_138283.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
NM_138283.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
NM_138283.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
NM_138283.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
NM_138283.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
NM_138283.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
NM_138283.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000006961.hg.1 |
NM_138283.1 |
Refseq |