Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05016060.hg.1 |
TC05001105.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05016060.hg.1 |
ENST00000296820.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05016060.hg.1 |
ENST00000310581.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05016060.hg.1 |
ENST00000334602.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05016060.hg.1 |
ENST00000508104.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05016060.hg.1 |
NM_001193376.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05016060.hg.1 |
NM_198253.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500169526.hg.1 |
ENST00000310581.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500169526.hg.1 |
ENST00000334602.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500169526.hg.1 |
ENST00000460137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500169526.hg.1 |
ENST00000484238.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500169526.hg.1 |
ENST00000508104.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500169526.hg.1 |
NM_001193376.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500169526.hg.1 |
NM_198253.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500169526.hg.1 |
ENST00000310581.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500169526.hg.1 |
ENST00000334602.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500169526.hg.1 |
ENST00000460137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500169526.hg.1 |
ENST00000484238.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500169526.hg.1 |
ENST00000508104.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500169526.hg.1 |
NM_001193376.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500169526.hg.1 |
NM_198253.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500169526.hg.1 |
TC0500009852.hg.1 |
Clariom D TC |