Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000479869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000479869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000479869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000479869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000479869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
ENST00000479869.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052594.hg.1 |
TC01003363.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052594.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052594.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052593.hg.1 |
TC01003363.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052593.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01052593.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221854.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221854.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221854.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221854.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221854.hg.1 |
TC0100016045.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000448509.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000479869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000479869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000479869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000479869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000479869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
ENST00000479869.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016045.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221853.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221853.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221853.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221853.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221853.hg.1 |
TC0100016045.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100221855.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221855.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221855.hg.1 |
ENST00000271532.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221855.hg.1 |
NM_031282.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221855.hg.1 |
TC0100016045.hg.1 |
Clariom D TC |