Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001018089.2 | NP_001018099.1 | 11 - 12 | 2386 | 122 | |
NM_024611.5 | NP_078887.2 | 11 - 12 | 2546 | 122 | ||
XM_005254661.1 | XP_005254718.1 | 12 - 13 | 2545 | 122 | ||
XM_005254662.1 | XP_005254719.1 | 12 - 13 | 2408 | 122 | ||
XM_011522009.1 | XP_011520311.1 | 11 - 12 | 2420 | 122 | ||
XM_011522010.1 | XP_011520312.1 | 11 - 12 | 2289 | 122 | ||
XM_011522011.2 | XP_011520313.1 | 10 - 11 | 2203 | 122 | ||
XM_017022569.1 | XP_016878058.1 | 12 - 13 | 2582 | 122 | ||
GenBank mRNA | AF502591.1 | - | 11 - 12 | 2489 | 122 | |
AK021958.1 | - | 2 - 3 | 540 | 122 | ||
AK055752.1 | - | 11 - 12 | 2489 | 122 | ||
AK095389.1 | - | 11 - 12 | 2370 | 122 | ||
AK127892.1 | - | 4 - 5 | 1371 | 122 | ||
AL832046.1 | - | 11 - 12 | 2513 | 122 | ||
BC013684.1 | - | 2 - 3 | 541 | 122 | ||
BC071685.1 | - | 2 - 3 | 435 | 122 | ||
BC107427.1 | - | 2 - 3 | 519 | 122 | ||
BC140894.1 | - | 11 - 12 | 2465 | 9021762 | 122 | |
BX647873.1 | - | 11 - 12 | 2372 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |