Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000242375.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000242375.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000242375.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000242375.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000242375.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000242375.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000242375.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000242375.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000242375.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000242375.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000242375.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000242375.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000242375.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000411726.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000411726.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000411726.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000411726.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000411726.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000411726.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000411726.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000411726.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000411726.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000411726.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000411726.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000411726.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000432161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000432161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000432161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000432161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000432161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000432161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000432161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000432161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000432161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000432161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000432161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000432161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000438242.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000438242.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000438242.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000438242.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000470851.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000470851.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
ENST00000470851.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_001190906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_001190906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_001190906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_001190906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_001190906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_001190906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_001190906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_001190906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_001190906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_001190906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_001190906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_001190906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_001190906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_001190906.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_001190907.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_001190907.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_001190907.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_001190907.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_001190907.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_001190907.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_001190907.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_001190907.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_001190907.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_001190907.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_001190907.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_001190907.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_001190907.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_001190907.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_005989.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_005989.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_005989.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_005989.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_005989.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_005989.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_005989.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_005989.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_005989.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_005989.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_005989.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_005989.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_005989.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_005989.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013466.hg.1 |
NM_005989.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013451.hg.1 |
TC07000858.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013451.hg.1 |
ENST00000242375.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013451.hg.1 |
ENST00000297463.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013451.hg.1 |
ENST00000432161.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013451.hg.1 |
ENST00000468877.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013451.hg.1 |
NM_001190907.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07013451.hg.1 |
NM_005989.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198963.hg.1 |
ENST00000242375.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198963.hg.1 |
ENST00000432161.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198963.hg.1 |
NM_001190907.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198963.hg.1 |
NM_005989.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198963.hg.1 |
ENST00000242375.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198963.hg.1 |
ENST00000432161.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700198963.hg.1 |
NM_001190907.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198963.hg.1 |
NM_005989.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700198963.hg.1 |
TC0700013466.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000242375.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000242375.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000242375.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000242375.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000242375.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000242375.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000242375.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000242375.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000242375.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000242375.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000242375.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000242375.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000242375.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000411726.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000411726.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000411726.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000411726.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000411726.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000411726.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000411726.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000411726.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000411726.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000411726.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000411726.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000411726.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000432161.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000432161.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000432161.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000432161.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000432161.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000432161.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000432161.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000432161.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000432161.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000432161.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000432161.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000432161.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000438242.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000438242.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000438242.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000438242.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000470851.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000470851.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
ENST00000470851.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_001190906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_001190906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_001190906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_001190906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_001190906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_001190906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_001190906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_001190906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_001190906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_001190906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_001190906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_001190906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_001190906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_001190906.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_001190907.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_001190907.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_001190907.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_001190907.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_001190907.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_001190907.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_001190907.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_001190907.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_001190907.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_001190907.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_001190907.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_001190907.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_001190907.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_001190907.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_005989.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_005989.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_005989.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_005989.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_005989.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_005989.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_005989.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_005989.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_005989.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_005989.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_005989.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_005989.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_005989.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_005989.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013466.hg.1 |
NM_005989.3 |
Refseq |