Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR18001954.hg.1 |
ENST00000282059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18001954.hg.1 |
ENST00000321319.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18001954.hg.1 |
NM_001008239.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18001954.hg.1 |
NM_145055.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18001954.hg.1 |
TC18000167.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR18001956.hg.1 |
ENST00000282059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18001956.hg.1 |
ENST00000321319.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18001956.hg.1 |
NM_001008239.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18001956.hg.1 |
NM_145055.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18001956.hg.1 |
TC18000167.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1800149534.hg.1 |
ENST00000615052.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800149534.hg.1 |
ENST00000619301.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800149534.hg.1 |
NM_001008239.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800149534.hg.1 |
NM_145055.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800149534.hg.1 |
TC1800007218.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800149534.hg.1 |
ENST00000615052.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800149534.hg.1 |
ENST00000619301.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800149534.hg.1 |
NM_001008239.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800149534.hg.1 |
NM_145055.3 |
Refseq |