Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAP1GAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015085.4 | NP_055900.4 | 6 - 7 | 362 | 72 | |
XM_005256542.2 | XP_005256599.1 | 5 - 6 | 754 | 72 | ||
XM_006721477.3 | XP_006721540.1 | 6 - 7 | 303 | 72 | ||
XM_011523738.1 | XP_011522040.1 | 7 - 8 | 415 | 72 | ||
XM_011523740.2 | XP_011522042.1 | 6 - 7 | 275 | 72 | ||
XM_011523741.2 | XP_011522043.1 | 6 - 7 | 451 | 72 | ||
XM_011523743.2 | XP_011522045.1 | 4 - 5 | 235 | 72 | ||
XM_017024370.1 | XP_016879859.1 | 6 - 7 | 340 | 72 | ||
XM_017024371.1 | XP_016879860.1 | 5 - 6 | 393 | 72 | ||
GenBank mRNA | AJ628446.1 | - | 6 - 7 | 362 | 72 | |
AK124640.1 | - | 6 - 7 | 451 | 72 | ||
BC140870.1 | - | 6 - 7 | 318 | 9021738 | 72 | |
BC140871.1 | - | 6 - 7 | 318 | 9021739 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |