Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011483.hg.1 |
ENST00000304883.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011483.hg.1 |
ENST00000304883.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011483.hg.1 |
ENST00000304883.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011483.hg.1 |
ENST00000304883.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011483.hg.1 |
ENST00000304883.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011483.hg.1 |
ENST00000304883.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011483.hg.1 |
ENST00000304883.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011483.hg.1 |
ENST00000304883.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011483.hg.1 |
ENST00000304883.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011483.hg.1 |
ENST00000304883.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011483.hg.1 |
NM_001059.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011483.hg.1 |
NM_001059.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011483.hg.1 |
NM_001059.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011483.hg.1 |
NM_001059.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011483.hg.1 |
NM_001059.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011483.hg.1 |
NM_001059.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011483.hg.1 |
NM_001059.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011483.hg.1 |
NM_001059.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011483.hg.1 |
NM_001059.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04020289.hg.1 |
TC04001439.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04020289.hg.1 |
ENST00000304883.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04020289.hg.1 |
NM_001059.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011483.hg.1 |
ENST00000304883.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011483.hg.1 |
ENST00000304883.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011483.hg.1 |
ENST00000304883.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011483.hg.1 |
ENST00000304883.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011483.hg.1 |
ENST00000304883.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011483.hg.1 |
ENST00000304883.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011483.hg.1 |
ENST00000304883.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011483.hg.1 |
ENST00000304883.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011483.hg.1 |
ENST00000304883.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011483.hg.1 |
ENST00000304883.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011483.hg.1 |
NM_001059.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011483.hg.1 |
NM_001059.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011483.hg.1 |
NM_001059.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011483.hg.1 |
NM_001059.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011483.hg.1 |
NM_001059.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011483.hg.1 |
NM_001059.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011483.hg.1 |
NM_001059.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011483.hg.1 |
NM_001059.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011483.hg.1 |
NM_001059.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400178828.hg.1 |
ENST00000304883.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400178828.hg.1 |
NM_001059.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400178828.hg.1 |
ENST00000304883.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400178828.hg.1 |
NM_001059.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400178828.hg.1 |
TC0400011483.hg.1 |
Clariom D TC |