Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006355.4 | NP_006346.1 | 6 - 7 | 1354 | 63 | |
XM_005248799.4 | XP_005248856.1 | 6 - 7 | 1083 | 63 | ||
XM_005248800.4 | XP_005248857.1 | 6 - 7 | 1224 | 63 | ||
XM_017010175.1 | XP_016865664.1 | 6 - 7 | 1243 | 63 | ||
GenBank mRNA | AB209906.1 | - | 4 - 5 | 3644 | 63 | |
AK313248.1 | - | 6 - 7 | 1241 | 63 | ||
BC026930.1 | - | 5 - 6 | 1200 | 3879491 | 63 | |
BX640949.1 | - | 6 - 7 | 1271 | 63 | ||
U90547.1 | - | 4 - 5 | 786 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |