Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
ENST00000220597.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
ENST00000220597.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
ENST00000220597.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
ENST00000220597.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
ENST00000220597.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
ENST00000220597.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
ENST00000220597.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
ENST00000220597.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
ENST00000220597.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
ENST00000220597.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
ENST00000220597.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
ENST00000220597.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
ENST00000220597.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
ENST00000220597.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
ENST00000220597.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
ENST00000220597.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
ENST00000220597.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
ENST00000220597.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
ENST00000220597.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
ENST00000519326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
ENST00000519326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
ENST00000519326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
ENST00000522811.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
ENST00000522811.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
ENST00000522811.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
ENST00000523463.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
ENST00000523463.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
NM_018440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
NM_018440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
NM_018440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
NM_018440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
NM_018440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
NM_018440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
NM_018440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
NM_018440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
NM_018440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
NM_018440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
NM_018440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
NM_018440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
NM_018440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
NM_018440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
NM_018440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
NM_018440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
NM_018440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
NM_018440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010926.hg.1 |
NM_018440.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08017563.hg.1 |
TC08001361.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08017563.hg.1 |
ENST00000220597.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08017563.hg.1 |
NM_018440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800174213.hg.1 |
ENST00000220597.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800174213.hg.1 |
NM_018440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800174213.hg.1 |
ENST00000220597.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800174213.hg.1 |
NM_018440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800174213.hg.1 |
TC0800010926.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
ENST00000220597.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
ENST00000220597.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
ENST00000220597.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
ENST00000220597.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
ENST00000220597.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
ENST00000220597.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
ENST00000220597.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
ENST00000220597.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
ENST00000220597.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
ENST00000220597.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
ENST00000220597.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
ENST00000220597.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
ENST00000220597.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
ENST00000220597.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
ENST00000220597.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
ENST00000220597.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
ENST00000220597.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
ENST00000220597.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
ENST00000220597.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
ENST00000519326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
ENST00000519326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
ENST00000519326.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
ENST00000522811.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
ENST00000522811.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
ENST00000522811.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
ENST00000523463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
ENST00000523463.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
NM_018440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
NM_018440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
NM_018440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
NM_018440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
NM_018440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
NM_018440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
NM_018440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
NM_018440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
NM_018440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
NM_018440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
NM_018440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
NM_018440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
NM_018440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
NM_018440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
NM_018440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
NM_018440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
NM_018440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
NM_018440.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010926.hg.1 |
NM_018440.3 |
Refseq |