Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01003203.hg.1 |
TC01000194.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01003203.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01003203.hg.1 |
ENST00000375799.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01003203.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000462455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000462455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000462455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000462455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000462455.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000477849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000477849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000477849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000477849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000477849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000477849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000477849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000477849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
ENST00000584345.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148802.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148802.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148802.hg.1 |
ENST00000462455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148802.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148802.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148802.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148802.hg.1 |
ENST00000462455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148802.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148802.hg.1 |
TC0100006994.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375793.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000375799.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000462455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000462455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000462455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000462455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000462455.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000477849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000477849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000477849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000477849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000477849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000477849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000477849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000477849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
ENST00000584345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006994.hg.1 |
NM_015164.3 |
Refseq |