Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19005822.hg.1 |
ENST00000270288.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19005822.hg.1 |
NM_001080436.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19005822.hg.1 |
TC19000445.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007810.hg.1 |
ENST00000585928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007810.hg.1 |
ENST00000585928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007810.hg.1 |
ENST00000585928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007810.hg.1 |
ENST00000585928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007810.hg.1 |
ENST00000585928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007810.hg.1 |
ENST00000585928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007810.hg.1 |
ENST00000585928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007810.hg.1 |
ENST00000585928.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007810.hg.1 |
ENST00000590071.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007810.hg.1 |
ENST00000590071.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007810.hg.1 |
ENST00000590071.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007810.hg.1 |
ENST00000590071.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007810.hg.1 |
ENST00000590071.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007810.hg.1 |
ENST00000590071.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007810.hg.1 |
ENST00000590071.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007810.hg.1 |
ENST00000590071.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007810.hg.1 |
ENST00000590071.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007810.hg.1 |
ENST00000590071.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007810.hg.1 |
ENST00000590071.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007810.hg.1 |
ENST00000590071.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007810.hg.1 |
NM_001080436.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007810.hg.1 |
NM_001080436.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007810.hg.1 |
NM_001080436.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007810.hg.1 |
NM_001080436.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007810.hg.1 |
NM_001080436.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007810.hg.1 |
NM_001080436.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007810.hg.1 |
NM_001080436.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007810.hg.1 |
NM_001080436.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007810.hg.1 |
NM_001080436.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007810.hg.1 |
NM_001080436.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900157490.hg.1 |
ENST00000585928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900157490.hg.1 |
ENST00000590071.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900157490.hg.1 |
NM_001080436.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900157490.hg.1 |
TC1900007810.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900157490.hg.1 |
ENST00000585928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900157490.hg.1 |
ENST00000590071.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900157490.hg.1 |
NM_001080436.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007810.hg.1 |
ENST00000585928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007810.hg.1 |
ENST00000585928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007810.hg.1 |
ENST00000585928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007810.hg.1 |
ENST00000585928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007810.hg.1 |
ENST00000585928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007810.hg.1 |
ENST00000585928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007810.hg.1 |
ENST00000585928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007810.hg.1 |
ENST00000585928.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007810.hg.1 |
ENST00000590071.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007810.hg.1 |
ENST00000590071.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007810.hg.1 |
ENST00000590071.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007810.hg.1 |
ENST00000590071.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007810.hg.1 |
ENST00000590071.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007810.hg.1 |
ENST00000590071.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007810.hg.1 |
ENST00000590071.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007810.hg.1 |
ENST00000590071.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007810.hg.1 |
ENST00000590071.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007810.hg.1 |
ENST00000590071.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007810.hg.1 |
ENST00000590071.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007810.hg.1 |
ENST00000590071.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007810.hg.1 |
NM_001080436.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007810.hg.1 |
NM_001080436.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007810.hg.1 |
NM_001080436.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007810.hg.1 |
NM_001080436.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007810.hg.1 |
NM_001080436.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007810.hg.1 |
NM_001080436.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007810.hg.1 |
NM_001080436.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007810.hg.1 |
NM_001080436.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007810.hg.1 |
NM_001080436.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007810.hg.1 |
NM_001080436.1 |
Refseq |