Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KNDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152643.6 | NP_689856.6 | 25 - 26 | 4526 | 62 | |
XM_017016858.1 | XP_016872347.1 | 25 - 26 | 4894 | 62 | ||
GenBank mRNA | AB000781.1 | - | 24 - 25 | 4422 | 62 | |
AK131205.1 | - | 6 - 7 | 807 | 62 | ||
AK294170.1 | - | 8 - 9 | 1120 | 62 | ||
BC047924.1 | - | 6 - 7 | 780 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |