Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03020385.hg.1 |
TC03001187.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03020385.hg.1 |
ENST00000273221.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03020385.hg.1 |
ENST00000435445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03020385.hg.1 |
ENST00000473088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03020385.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03020385.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03020387.hg.1 |
TC03001187.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03020387.hg.1 |
ENST00000273221.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03020387.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000273221.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000273221.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000273221.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000273221.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000273221.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000273221.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000273221.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000273221.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000273221.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000273221.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000273221.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000273221.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000273221.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000273221.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000273221.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000273221.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000273221.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000473088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000473088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000473088.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000474467.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000474467.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000613206.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000613206.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000613206.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000613206.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000613206.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000613206.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000613206.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000613206.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000613206.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000613206.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000613206.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000613206.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000613206.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000613206.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000613206.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000613206.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000618604.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000618604.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000618604.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000618604.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000618604.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000618604.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000618604.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000618604.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000618604.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000618604.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000618604.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000618604.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000618604.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000618604.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000618604.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
ENST00000618604.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010315.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300175295.hg.1 |
ENST00000273221.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300175295.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300175295.hg.1 |
ENST00000273221.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300175295.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300175295.hg.1 |
TC0300010315.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300175296.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300175296.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300175296.hg.1 |
TC0300010315.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300175291.hg.1 |
ENST00000273221.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300175291.hg.1 |
ENST00000473088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300175291.hg.1 |
ENST00000613206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300175291.hg.1 |
ENST00000618604.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300175291.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300175291.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300175291.hg.1 |
ENST00000273221.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300175291.hg.1 |
ENST00000473088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300175291.hg.1 |
ENST00000613206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300175291.hg.1 |
ENST00000618604.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300175291.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300175291.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300175291.hg.1 |
TC0300010315.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000273221.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000273221.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000273221.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000273221.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000273221.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000273221.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000273221.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000273221.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000273221.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000273221.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000273221.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000273221.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000273221.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000273221.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000273221.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000273221.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000273221.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000473088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000473088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000473088.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000474467.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000474467.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000613206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000613206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000613206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000613206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000613206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000613206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000613206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000613206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000613206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000613206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000613206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000613206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000613206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000613206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000613206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000613206.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000618604.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000618604.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000618604.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000618604.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000618604.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000618604.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000618604.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000618604.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000618604.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000618604.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000618604.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000618604.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000618604.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000618604.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000618604.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
ENST00000618604.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
NM_001134382.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010315.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300175293.hg.1 |
ENST00000273221.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300175293.hg.1 |
ENST00000474467.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300175293.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300175293.hg.1 |
ENST00000273221.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300175293.hg.1 |
ENST00000474467.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300175293.hg.1 |
NM_014869.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300175293.hg.1 |
TC0300010315.hg.1 |
Clariom D TC |