Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQSEC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134382.2 | NP_001127854.1 | 8 - 9 | 2292 | 75 | |
NM_014869.6 | NP_055684.3 | 8 - 9 | 2478 | 75 | ||
XM_011534304.2 | XP_011532606.1 | 10 - 11 | 2529 | 75 | ||
XM_011534305.2 | XP_011532607.1 | 8 - 9 | 2278 | 75 | ||
XM_011534306.2 | XP_011532608.1 | 8 - 9 | 2684 | 75 | ||
XM_011534307.2 | XP_011532609.1 | 9 - 10 | 2630 | 75 | ||
XM_011534308.2 | XP_011532610.1 | 8 - 9 | 2473 | 75 | ||
XM_011534310.2 | XP_011532612.1 | 10 - 11 | 2529 | 75 | ||
XM_011534311.2 | XP_011532613.1 | 8 - 9 | 2641 | 75 | ||
XM_011534312.2 | XP_011532614.1 | 8 - 9 | 2269 | 75 | ||
XM_011534313.2 | XP_011532615.1 | 8 - 9 | 2488 | 75 | ||
XM_011534314.2 | XP_011532616.1 | 8 - 9 | 2224 | 75 | ||
XM_011534315.2 | XP_011532617.1 | 8 - 9 | 2227 | 75 | ||
XM_017007582.1 | XP_016863071.1 | 9 - 10 | 2484 | 75 | ||
GenBank mRNA | AB018306.1 | - | 7 - 8 | 1971 | 75 | |
AK294525.1 | - | 8 - 9 | 2292 | 75 | ||
AY653734.1 | - | 8 - 9 | 2240 | 75 | ||
BC010267.1 | - | 8 - 9 | 2232 | 2900367 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |