Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022824.hg.1 |
ENST00000262013.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022824.hg.1 |
ENST00000357122.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022824.hg.1 |
ENST00000357791.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022824.hg.1 |
ENST00000357804.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022824.hg.1 |
ENST00000376407.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022824.hg.1 |
ENST00000505279.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022824.hg.1 |
ENST00000510283.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022824.hg.1 |
ENST00000535445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022824.hg.1 |
NM_001130527.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022824.hg.1 |
NM_001130528.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022824.hg.1 |
NM_001251971.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022824.hg.1 |
NM_003971.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17022824.hg.1 |
TC17001700.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700189557.hg.1 |
ENST00000262013.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189557.hg.1 |
ENST00000357122.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189557.hg.1 |
ENST00000505279.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189557.hg.1 |
ENST00000510283.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189557.hg.1 |
ENST00000618113.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189557.hg.1 |
NM_001130527.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700189557.hg.1 |
NM_001130528.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700189557.hg.1 |
NM_001251971.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700189557.hg.1 |
NM_003971.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700189557.hg.1 |
ENST00000262013.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189557.hg.1 |
ENST00000357122.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189557.hg.1 |
ENST00000505279.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189557.hg.1 |
ENST00000510283.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189557.hg.1 |
ENST00000618113.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700189557.hg.1 |
NM_001130527.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700189557.hg.1 |
NM_001130528.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700189557.hg.1 |
NM_001251971.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700189557.hg.1 |
NM_003971.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700189557.hg.1 |
TC1700011128.hg.1 |
Clariom D TC |