Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006349.hg.1 |
TC09000572.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006349.hg.1 |
ENST00000374210.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006349.hg.1 |
ENST00000374212.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006349.hg.1 |
NM_001859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008483.hg.1 |
ENST00000374212.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008483.hg.1 |
ENST00000374212.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008483.hg.1 |
ENST00000374212.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008483.hg.1 |
ENST00000374212.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008483.hg.1 |
ENST00000374212.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008483.hg.1 |
ENST00000374212.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008483.hg.1 |
ENST00000374212.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008483.hg.1 |
ENST00000374212.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008483.hg.1 |
ENST00000374212.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008483.hg.1 |
ENST00000374212.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008483.hg.1 |
ENST00000496650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008483.hg.1 |
ENST00000496650.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008483.hg.1 |
NM_001859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008483.hg.1 |
NM_001859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008483.hg.1 |
NM_001859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008483.hg.1 |
NM_001859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008483.hg.1 |
NM_001859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008483.hg.1 |
NM_001859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008483.hg.1 |
NM_001859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008483.hg.1 |
NM_001859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008483.hg.1 |
NM_001859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008483.hg.1 |
NM_001859.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008483.hg.1 |
NM_001859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008483.hg.1 |
ENST00000374212.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008483.hg.1 |
ENST00000374212.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008483.hg.1 |
ENST00000374212.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008483.hg.1 |
ENST00000374212.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008483.hg.1 |
ENST00000374212.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008483.hg.1 |
ENST00000374212.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008483.hg.1 |
ENST00000374212.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008483.hg.1 |
ENST00000374212.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008483.hg.1 |
ENST00000374212.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008483.hg.1 |
ENST00000374212.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008483.hg.1 |
ENST00000496650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008483.hg.1 |
ENST00000496650.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008483.hg.1 |
NM_001859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008483.hg.1 |
NM_001859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008483.hg.1 |
NM_001859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008483.hg.1 |
NM_001859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008483.hg.1 |
NM_001859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008483.hg.1 |
NM_001859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008483.hg.1 |
NM_001859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008483.hg.1 |
NM_001859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008483.hg.1 |
NM_001859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008483.hg.1 |
NM_001859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008483.hg.1 |
NM_001859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900156627.hg.1 |
ENST00000374212.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156627.hg.1 |
NM_001859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900156627.hg.1 |
ENST00000374212.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900156627.hg.1 |
NM_001859.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900156627.hg.1 |
TC0900008483.hg.1 |
Clariom D TC |