Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009183.hg.1 |
TC09000731.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009183.hg.1 |
ENST00000372398.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009183.hg.1 |
ENST00000458469.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009183.hg.1 |
NM_001128826.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009183.hg.1 |
NM_014286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009184.hg.1 |
TC09000731.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009184.hg.1 |
ENST00000372398.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009184.hg.1 |
ENST00000458469.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009184.hg.1 |
NM_001128826.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009184.hg.1 |
NM_014286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
ENST00000372398.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
ENST00000372398.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
ENST00000372398.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
ENST00000372398.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
ENST00000372398.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
ENST00000372398.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
ENST00000372398.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
ENST00000372398.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
ENST00000372398.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
ENST00000372398.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
ENST00000372398.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
ENST00000372398.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
ENST00000493042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
ENST00000493042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
ENST00000493042.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
ENST00000630865.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
ENST00000630865.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
ENST00000630865.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
ENST00000630865.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
ENST00000630865.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
ENST00000630865.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
ENST00000630865.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
ENST00000630865.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
ENST00000630865.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
NM_001128826.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
NM_001128826.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
NM_001128826.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
NM_001128826.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
NM_001128826.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
NM_001128826.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
NM_001128826.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
NM_001128826.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
NM_001128826.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
NM_001128826.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
NM_014286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
NM_014286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
NM_014286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
NM_014286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
NM_014286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
NM_014286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
NM_014286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
NM_014286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
NM_014286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
NM_014286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
NM_014286.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008949.hg.1 |
NM_014286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900160687.hg.1 |
ENST00000372398.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900160687.hg.1 |
ENST00000630865.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900160687.hg.1 |
NM_001128826.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900160687.hg.1 |
NM_014286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900160687.hg.1 |
ENST00000372398.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900160687.hg.1 |
ENST00000630865.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900160687.hg.1 |
NM_001128826.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900160687.hg.1 |
NM_014286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900160687.hg.1 |
TC0900008949.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900160688.hg.1 |
ENST00000372398.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900160688.hg.1 |
ENST00000630865.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900160688.hg.1 |
NM_001128826.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900160688.hg.1 |
NM_014286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900160688.hg.1 |
ENST00000372398.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900160688.hg.1 |
ENST00000630865.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900160688.hg.1 |
NM_001128826.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900160688.hg.1 |
NM_014286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900160688.hg.1 |
TC0900008949.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
ENST00000372398.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
ENST00000372398.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
ENST00000372398.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
ENST00000372398.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
ENST00000372398.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
ENST00000372398.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
ENST00000372398.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
ENST00000372398.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
ENST00000372398.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
ENST00000372398.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
ENST00000372398.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
ENST00000372398.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
ENST00000493042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
ENST00000493042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
ENST00000493042.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
ENST00000630865.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
ENST00000630865.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
ENST00000630865.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
ENST00000630865.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
ENST00000630865.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
ENST00000630865.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
ENST00000630865.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
ENST00000630865.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
ENST00000630865.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
NM_001128826.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
NM_001128826.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
NM_001128826.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
NM_001128826.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
NM_001128826.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
NM_001128826.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
NM_001128826.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
NM_001128826.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
NM_001128826.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
NM_001128826.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
NM_014286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
NM_014286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
NM_014286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
NM_014286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
NM_014286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
NM_014286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
NM_014286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
NM_014286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
NM_014286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
NM_014286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
NM_014286.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008949.hg.1 |
NM_014286.3 |
Refseq |