Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
ENST00000258455.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
ENST00000258455.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
ENST00000258455.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
ENST00000258455.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
ENST00000258455.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
ENST00000258455.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
ENST00000258455.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
ENST00000258455.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
ENST00000258455.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
ENST00000258455.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
ENST00000258455.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
ENST00000258455.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
ENST00000258455.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
ENST00000258455.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
ENST00000258455.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
ENST00000258455.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
ENST00000258455.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
ENST00000413583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
ENST00000413583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
ENST00000413583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
ENST00000413583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
ENST00000413583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
ENST00000413583.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
ENST00000472220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
ENST00000472220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
ENST00000472220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
ENST00000472220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
ENST00000472220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
ENST00000472220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
ENST00000472220.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
NM_182640.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
NM_182640.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
NM_182640.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
NM_182640.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
NM_182640.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
NM_182640.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
NM_182640.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
NM_182640.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
NM_182640.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
NM_182640.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
NM_182640.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
NM_182640.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
NM_182640.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
NM_182640.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
NM_182640.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200008724.hg.1 |
NM_182640.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010081.hg.1 |
TC02000642.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010081.hg.1 |
ENST00000258455.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010081.hg.1 |
NM_182640.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010079.hg.1 |
TC02000642.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010079.hg.1 |
ENST00000258455.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010079.hg.1 |
NM_182640.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010080.hg.1 |
TC02000642.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010080.hg.1 |
ENST00000258455.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02010080.hg.1 |
NM_182640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200160937.hg.1 |
ENST00000258455.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200160937.hg.1 |
NM_182640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200160937.hg.1 |
ENST00000258455.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200160937.hg.1 |
NM_182640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200160937.hg.1 |
TC0200008724.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200160939.hg.1 |
ENST00000258455.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200160939.hg.1 |
NM_182640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200160939.hg.1 |
ENST00000258455.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200160939.hg.1 |
NM_182640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200160939.hg.1 |
TC0200008724.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
ENST00000258455.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
ENST00000258455.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
ENST00000258455.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
ENST00000258455.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
ENST00000258455.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
ENST00000258455.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
ENST00000258455.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
ENST00000258455.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
ENST00000258455.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
ENST00000258455.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
ENST00000258455.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
ENST00000258455.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
ENST00000258455.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
ENST00000258455.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
ENST00000258455.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
ENST00000258455.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
ENST00000258455.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
ENST00000413583.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
ENST00000413583.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
ENST00000413583.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
ENST00000413583.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
ENST00000413583.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
ENST00000413583.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
ENST00000472220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
ENST00000472220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
ENST00000472220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
ENST00000472220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
ENST00000472220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
ENST00000472220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
ENST00000472220.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
NM_182640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
NM_182640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
NM_182640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
NM_182640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
NM_182640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
NM_182640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
NM_182640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
NM_182640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
NM_182640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
NM_182640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
NM_182640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
NM_182640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
NM_182640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
NM_182640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
NM_182640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200008724.hg.1 |
NM_182640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200160940.hg.1 |
ENST00000258455.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200160940.hg.1 |
NM_182640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200160940.hg.1 |
ENST00000258455.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200160940.hg.1 |
NM_182640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200160940.hg.1 |
TC0200008724.hg.1 |
Clariom D TC |