Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006525.hg.1 |
ENST00000409293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006525.hg.1 |
ENST00000409293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006525.hg.1 |
ENST00000409293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006525.hg.1 |
ENST00000409293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006525.hg.1 |
ENST00000409293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006525.hg.1 |
ENST00000409293.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006525.hg.1 |
ENST00000469144.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006525.hg.1 |
ENST00000469144.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006525.hg.1 |
ENST00000469144.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006525.hg.1 |
ENST00000469144.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006525.hg.1 |
ENST00000485191.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006525.hg.1 |
ENST00000485191.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006525.hg.1 |
ENST00000485191.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006525.hg.1 |
ENST00000485191.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006525.hg.1 |
ENST00000485191.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006525.hg.1 |
ENST00000485191.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006525.hg.1 |
ENST00000485191.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006525.hg.1 |
ENST00000585716.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006525.hg.1 |
ENST00000585716.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006525.hg.1 |
ENST00000585716.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006525.hg.1 |
ENST00000585716.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006525.hg.1 |
ENST00000590269.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006525.hg.1 |
ENST00000590269.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006525.hg.1 |
ENST00000590269.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006525.hg.1 |
NM_017914.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006525.hg.1 |
NM_017914.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006525.hg.1 |
NM_017914.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006525.hg.1 |
NM_017914.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006525.hg.1 |
NM_017914.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006525.hg.1 |
ENST00000409293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006525.hg.1 |
ENST00000409293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006525.hg.1 |
ENST00000409293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006525.hg.1 |
ENST00000409293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006525.hg.1 |
ENST00000409293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006525.hg.1 |
ENST00000409293.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006525.hg.1 |
ENST00000469144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006525.hg.1 |
ENST00000469144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006525.hg.1 |
ENST00000469144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006525.hg.1 |
ENST00000469144.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006525.hg.1 |
ENST00000485191.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006525.hg.1 |
ENST00000485191.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006525.hg.1 |
ENST00000485191.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006525.hg.1 |
ENST00000485191.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006525.hg.1 |
ENST00000485191.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006525.hg.1 |
ENST00000485191.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006525.hg.1 |
ENST00000485191.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006525.hg.1 |
ENST00000585716.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006525.hg.1 |
ENST00000585716.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006525.hg.1 |
ENST00000585716.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006525.hg.1 |
ENST00000585716.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006525.hg.1 |
ENST00000590269.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006525.hg.1 |
ENST00000590269.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006525.hg.1 |
ENST00000590269.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006525.hg.1 |
NM_017914.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006525.hg.1 |
NM_017914.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006525.hg.1 |
NM_017914.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006525.hg.1 |
NM_017914.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006525.hg.1 |
NM_017914.3 |
Refseq |