Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02015991.hg.1 |
TC02001074.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02015991.hg.1 |
ENST00000234453.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02015991.hg.1 |
NM_019091.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
ENST00000234453.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
ENST00000234453.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
ENST00000234453.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
ENST00000234453.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
ENST00000234453.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
ENST00000234453.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
ENST00000234453.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
ENST00000234453.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
ENST00000234453.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
ENST00000234453.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
ENST00000234453.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
ENST00000234453.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
ENST00000234453.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
ENST00000234453.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
ENST00000234453.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
ENST00000234453.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
ENST00000421187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
ENST00000421187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
ENST00000421187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
ENST00000421187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
ENST00000421187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
ENST00000421187.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
ENST00000453653.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
ENST00000453653.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
ENST00000453653.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
ENST00000453653.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
ENST00000453653.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
ENST00000453653.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
ENST00000453653.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
ENST00000453653.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
ENST00000461474.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
ENST00000461474.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
ENST00000461474.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
ENST00000461474.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
NM_019091.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
NM_019091.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
NM_019091.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
NM_019091.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
NM_019091.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
NM_019091.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
NM_019091.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
NM_019091.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
NM_019091.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
NM_019091.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
NM_019091.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
NM_019091.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
NM_019091.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
NM_019091.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010119.hg.1 |
NM_019091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200170969.hg.1 |
ENST00000234453.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200170969.hg.1 |
ENST00000453653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200170969.hg.1 |
NM_019091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200170969.hg.1 |
ENST00000234453.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200170969.hg.1 |
ENST00000453653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200170969.hg.1 |
NM_019091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200170969.hg.1 |
TC0200010119.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
ENST00000234453.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
ENST00000234453.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
ENST00000234453.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
ENST00000234453.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
ENST00000234453.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
ENST00000234453.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
ENST00000234453.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
ENST00000234453.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
ENST00000234453.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
ENST00000234453.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
ENST00000234453.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
ENST00000234453.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
ENST00000234453.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
ENST00000234453.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
ENST00000234453.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
ENST00000234453.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
ENST00000421187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
ENST00000421187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
ENST00000421187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
ENST00000421187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
ENST00000421187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
ENST00000421187.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
ENST00000453653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
ENST00000453653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
ENST00000453653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
ENST00000453653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
ENST00000453653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
ENST00000453653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
ENST00000453653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
ENST00000453653.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
ENST00000461474.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
ENST00000461474.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
ENST00000461474.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
ENST00000461474.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
NM_019091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
NM_019091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
NM_019091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
NM_019091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
NM_019091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
NM_019091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
NM_019091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
NM_019091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
NM_019091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
NM_019091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
NM_019091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
NM_019091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
NM_019091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
NM_019091.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010119.hg.1 |
NM_019091.3 |
Refseq |