Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08017373.hg.1 |
TC08001350.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08017373.hg.1 |
ENST00000263851.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08017373.hg.1 |
ENST00000379114.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08017373.hg.1 |
ENST00000519833.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08017373.hg.1 |
ENST00000520269.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08017373.hg.1 |
NM_000880.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08017373.hg.1 |
NM_001199886.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800173867.hg.1 |
ENST00000263851.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173867.hg.1 |
ENST00000518982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173867.hg.1 |
ENST00000519833.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173867.hg.1 |
ENST00000520215.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173867.hg.1 |
ENST00000520269.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173867.hg.1 |
ENST00000520317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173867.hg.1 |
ENST00000523959.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173867.hg.1 |
NM_000880.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800173867.hg.1 |
NM_001199886.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800173867.hg.1 |
ENST00000263851.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173867.hg.1 |
ENST00000518982.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173867.hg.1 |
ENST00000519833.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173867.hg.1 |
ENST00000520215.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173867.hg.1 |
ENST00000520269.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173867.hg.1 |
ENST00000520317.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173867.hg.1 |
ENST00000523959.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800173867.hg.1 |
NM_000880.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800173867.hg.1 |
NM_001199886.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800173867.hg.1 |
TC0800010880.hg.1 |
Clariom D TC |