Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS6KC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287219.1 | NP_001274148.1 | 1 - 2 | 275 | 117 | |
XM_005273096.1 | XP_005273153.1 | 1 - 2 | 256 | 117 | ||
XM_017001027.1 | XP_016856516.1 | 1 - 2 | 256 | 117 | ||
XM_017001028.1 | XP_016856517.1 | 1 - 2 | 256 | 117 | ||
XM_017001029.1 | XP_016856518.1 | 1 - 2 | 256 | 117 | ||
XM_017001030.1 | XP_016856519.1 | 1 - 2 | 256 | 117 | ||
XM_017001031.1 | XP_016856520.1 | 1 - 2 | 256 | 117 | ||
XM_017001033.1 | XP_016856522.1 | 1 - 2 | 256 | 117 | ||
XM_017001034.1 | XP_016856523.1 | 1 - 2 | 256 | 117 | ||
GenBank mRNA | AK299298.1 | - | 1 - 2 | 261 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Kinase Homo sapiens Probe spans exons |