Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016492.hg.1 |
TC08001282.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016492.hg.1 |
ENST00000379419.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016492.hg.1 |
ENST00000396642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016492.hg.1 |
ENST00000401827.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016492.hg.1 |
ENST00000518667.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016492.hg.1 |
NM_001242318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016492.hg.1 |
NM_002603.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016496.hg.1 |
TC08001282.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016496.hg.1 |
ENST00000379419.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016496.hg.1 |
ENST00000396642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016496.hg.1 |
ENST00000401827.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016496.hg.1 |
ENST00000518667.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016496.hg.1 |
NM_001242318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016496.hg.1 |
NM_002603.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016493.hg.1 |
TC08001282.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016493.hg.1 |
ENST00000379419.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016493.hg.1 |
ENST00000396642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016493.hg.1 |
ENST00000401827.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016493.hg.1 |
ENST00000518667.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016493.hg.1 |
NM_001242318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016493.hg.1 |
NM_002603.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016494.hg.1 |
TC08001282.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016494.hg.1 |
ENST00000379419.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016494.hg.1 |
ENST00000396642.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016494.hg.1 |
ENST00000401827.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016494.hg.1 |
ENST00000518667.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016494.hg.1 |
NM_001242318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016494.hg.1 |
NM_002603.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172405.hg.1 |
ENST00000379419.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172405.hg.1 |
ENST00000396642.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172405.hg.1 |
ENST00000401827.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172405.hg.1 |
ENST00000518667.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172405.hg.1 |
ENST00000519231.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172405.hg.1 |
ENST00000519626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172405.hg.1 |
ENST00000523253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172405.hg.1 |
NM_001242318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172405.hg.1 |
NM_002603.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172405.hg.1 |
ENST00000379419.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172405.hg.1 |
ENST00000396642.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172405.hg.1 |
ENST00000401827.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172405.hg.1 |
ENST00000518667.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172405.hg.1 |
ENST00000519231.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172405.hg.1 |
ENST00000519626.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172405.hg.1 |
ENST00000523253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172405.hg.1 |
NM_001242318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172405.hg.1 |
NM_002603.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172405.hg.1 |
TC0800010667.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800172401.hg.1 |
ENST00000379419.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172401.hg.1 |
ENST00000396642.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172401.hg.1 |
ENST00000401827.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172401.hg.1 |
ENST00000518667.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172401.hg.1 |
NM_001242318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172401.hg.1 |
NM_002603.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172401.hg.1 |
ENST00000379419.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172401.hg.1 |
ENST00000396642.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172401.hg.1 |
ENST00000401827.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172401.hg.1 |
ENST00000518667.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172401.hg.1 |
NM_001242318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172401.hg.1 |
NM_002603.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172401.hg.1 |
TC0800010667.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800172402.hg.1 |
ENST00000379419.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172402.hg.1 |
ENST00000396642.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172402.hg.1 |
ENST00000401827.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172402.hg.1 |
ENST00000518667.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172402.hg.1 |
NM_001242318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172402.hg.1 |
NM_002603.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172402.hg.1 |
ENST00000379419.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172402.hg.1 |
ENST00000396642.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172402.hg.1 |
ENST00000401827.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172402.hg.1 |
ENST00000518667.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172402.hg.1 |
NM_001242318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172402.hg.1 |
NM_002603.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172402.hg.1 |
TC0800010667.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800172403.hg.1 |
ENST00000379419.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172403.hg.1 |
ENST00000396642.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172403.hg.1 |
ENST00000401827.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172403.hg.1 |
ENST00000518667.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172403.hg.1 |
ENST00000519231.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172403.hg.1 |
NM_001242318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172403.hg.1 |
NM_002603.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172403.hg.1 |
ENST00000379419.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172403.hg.1 |
ENST00000396642.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172403.hg.1 |
ENST00000401827.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172403.hg.1 |
ENST00000518667.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172403.hg.1 |
ENST00000519231.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172403.hg.1 |
NM_001242318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172403.hg.1 |
NM_002603.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172403.hg.1 |
TC0800010667.hg.1 |
Clariom D TC |