Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03000231.hg.1 |
TC03000011.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03000231.hg.1 |
ENST00000319331.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03000231.hg.1 |
NM_020873.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03000221.hg.1 |
TC03000011.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03000221.hg.1 |
ENST00000319331.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03000221.hg.1 |
NM_020873.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006465.hg.1 |
ENST00000319331.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006465.hg.1 |
ENST00000319331.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006465.hg.1 |
ENST00000319331.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006465.hg.1 |
ENST00000319331.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006465.hg.1 |
ENST00000319331.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006465.hg.1 |
ENST00000319331.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006465.hg.1 |
ENST00000319331.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006465.hg.1 |
ENST00000319331.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006465.hg.1 |
ENST00000472075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006465.hg.1 |
ENST00000472075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006465.hg.1 |
ENST00000472075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006465.hg.1 |
ENST00000472075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006465.hg.1 |
ENST00000496115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006465.hg.1 |
ENST00000496115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006465.hg.1 |
ENST00000496115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006465.hg.1 |
NM_020873.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006465.hg.1 |
NM_020873.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006465.hg.1 |
NM_020873.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006465.hg.1 |
NM_020873.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006465.hg.1 |
NM_020873.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006465.hg.1 |
NM_020873.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006465.hg.1 |
NM_020873.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300006465.hg.1 |
NM_020873.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03000224.hg.1 |
TC03000011.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03000224.hg.1 |
ENST00000319331.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03000224.hg.1 |
NM_020873.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013924.hg.1 |
ENST00000469888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013924.hg.1 |
ENST00000469888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013924.hg.1 |
ENST00000469888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013924.hg.1 |
ENST00000469888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013924.hg.1 |
ENST00000470751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013924.hg.1 |
ENST00000470751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013924.hg.1 |
ENST00000470751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013924.hg.1 |
ENST00000470751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013924.hg.1 |
ENST00000470751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013924.hg.1 |
ENST00000470751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013924.hg.1 |
ENST00000470751.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013924.hg.1 |
ENST00000484993.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013924.hg.1 |
ENST00000484993.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013924.hg.1 |
ENST00000484993.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03000230.hg.1 |
TC03000011.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03000230.hg.1 |
ENST00000319331.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03000230.hg.1 |
ENST00000496115.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03000230.hg.1 |
NM_020873.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03000232.hg.1 |
TC03000011.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03000232.hg.1 |
ENST00000319331.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03000232.hg.1 |
NM_020873.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03000220.hg.1 |
TC03000011.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03000220.hg.1 |
ENST00000319331.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03000220.hg.1 |
NM_020873.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300144340.hg.1 |
ENST00000319331.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300144340.hg.1 |
ENST00000472075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300144340.hg.1 |
NM_020873.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300144340.hg.1 |
ENST00000319331.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300144340.hg.1 |
ENST00000472075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300144340.hg.1 |
NM_020873.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300144340.hg.1 |
TC0300006465.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300144338.hg.1 |
ENST00000319331.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300144338.hg.1 |
NM_020873.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300144338.hg.1 |
ENST00000319331.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300144338.hg.1 |
NM_020873.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300144338.hg.1 |
TC0300006465.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300006465.hg.1 |
ENST00000319331.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006465.hg.1 |
ENST00000319331.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006465.hg.1 |
ENST00000319331.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006465.hg.1 |
ENST00000319331.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006465.hg.1 |
ENST00000319331.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006465.hg.1 |
ENST00000319331.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006465.hg.1 |
ENST00000319331.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006465.hg.1 |
ENST00000319331.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006465.hg.1 |
ENST00000472075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006465.hg.1 |
ENST00000472075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006465.hg.1 |
ENST00000472075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006465.hg.1 |
ENST00000472075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006465.hg.1 |
ENST00000496115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006465.hg.1 |
ENST00000496115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006465.hg.1 |
ENST00000496115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300006465.hg.1 |
NM_020873.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006465.hg.1 |
NM_020873.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006465.hg.1 |
NM_020873.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006465.hg.1 |
NM_020873.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006465.hg.1 |
NM_020873.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006465.hg.1 |
NM_020873.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006465.hg.1 |
NM_020873.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300006465.hg.1 |
NM_020873.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300144339.hg.1 |
ENST00000319331.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300144339.hg.1 |
ENST00000472075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300144339.hg.1 |
NM_020873.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300144339.hg.1 |
ENST00000319331.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300144339.hg.1 |
ENST00000472075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300144339.hg.1 |
NM_020873.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300144339.hg.1 |
TC0300006465.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300144345.hg.1 |
ENST00000319331.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300144345.hg.1 |
NM_020873.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300144345.hg.1 |
ENST00000319331.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300144345.hg.1 |
NM_020873.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300144345.hg.1 |
TC0300006465.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300144346.hg.1 |
ENST00000319331.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300144346.hg.1 |
NM_020873.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300144346.hg.1 |
ENST00000319331.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300144346.hg.1 |
NM_020873.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300144346.hg.1 |
TC0300006465.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300144344.hg.1 |
ENST00000319331.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300144344.hg.1 |
ENST00000496115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300144344.hg.1 |
NM_020873.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300144344.hg.1 |
ENST00000319331.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300144344.hg.1 |
ENST00000496115.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300144344.hg.1 |
NM_020873.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300144344.hg.1 |
TC0300006465.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300013924.hg.1 |
ENST00000469888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013924.hg.1 |
ENST00000469888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013924.hg.1 |
ENST00000469888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013924.hg.1 |
ENST00000469888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013924.hg.1 |
ENST00000470751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013924.hg.1 |
ENST00000470751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013924.hg.1 |
ENST00000470751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013924.hg.1 |
ENST00000470751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013924.hg.1 |
ENST00000470751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013924.hg.1 |
ENST00000470751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013924.hg.1 |
ENST00000470751.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013924.hg.1 |
ENST00000484993.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013924.hg.1 |
ENST00000484993.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013924.hg.1 |
ENST00000484993.1 |
Ensembl |