Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002207.2 | NP_002198.2 | 13 - 14 | 1420 | 74 | |
GenBank mRNA | BC030198.1 | - | 13 - 14 | 1627 | 5199296 | 74 |
D25303.1 | - | 14 - 15 | 1418 | 74 | ||
L24158.1 | - | 13 - 14 | 1262 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |