Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17023068.hg.1 |
ENST00000258963.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17023068.hg.1 |
NM_007146.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17023068.hg.1 |
TC17001722.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
ENST00000258963.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
ENST00000258963.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
ENST00000258963.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
ENST00000258963.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
ENST00000258963.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
ENST00000258963.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
ENST00000258963.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
ENST00000581208.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
ENST00000581208.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
ENST00000581208.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
ENST00000581208.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
ENST00000581208.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
ENST00000581208.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
ENST00000581208.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
ENST00000581208.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
ENST00000581208.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
ENST00000581208.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
ENST00000583932.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
ENST00000583932.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
ENST00000583932.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
ENST00000583932.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
ENST00000583932.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
ENST00000584396.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
ENST00000584396.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
ENST00000584396.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
ENST00000584396.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
ENST00000584396.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
ENST00000584396.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
ENST00000584396.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
ENST00000584396.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
ENST00000584396.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
NM_007146.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
NM_007146.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
NM_007146.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
NM_007146.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
NM_007146.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
NM_007146.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
NM_007146.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
NM_007146.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
NM_007146.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
NM_007146.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
NM_007146.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011237.hg.1 |
NM_007146.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700190236.hg.1 |
ENST00000581208.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190236.hg.1 |
ENST00000584396.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190236.hg.1 |
NM_007146.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700190236.hg.1 |
ENST00000581208.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190236.hg.1 |
ENST00000584396.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190236.hg.1 |
NM_007146.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700190236.hg.1 |
TC1700011237.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700190234.hg.1 |
ENST00000258963.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190234.hg.1 |
ENST00000581208.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190234.hg.1 |
ENST00000584396.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190234.hg.1 |
NM_007146.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700190234.hg.1 |
ENST00000258963.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190234.hg.1 |
ENST00000581208.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190234.hg.1 |
ENST00000584396.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190234.hg.1 |
NM_007146.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700190234.hg.1 |
TC1700011237.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
ENST00000258963.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
ENST00000258963.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
ENST00000258963.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
ENST00000258963.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
ENST00000258963.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
ENST00000258963.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
ENST00000258963.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
ENST00000581208.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
ENST00000581208.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
ENST00000581208.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
ENST00000581208.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
ENST00000581208.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
ENST00000581208.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
ENST00000581208.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
ENST00000581208.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
ENST00000581208.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
ENST00000581208.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
ENST00000583932.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
ENST00000583932.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
ENST00000583932.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
ENST00000583932.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
ENST00000583932.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
ENST00000584396.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
ENST00000584396.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
ENST00000584396.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
ENST00000584396.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
ENST00000584396.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
ENST00000584396.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
ENST00000584396.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
ENST00000584396.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
ENST00000584396.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
NM_007146.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
NM_007146.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
NM_007146.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
NM_007146.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
NM_007146.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
NM_007146.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
NM_007146.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
NM_007146.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
NM_007146.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
NM_007146.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
NM_007146.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700011237.hg.1 |
NM_007146.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700190235.hg.1 |
ENST00000258963.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190235.hg.1 |
ENST00000581208.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190235.hg.1 |
ENST00000584396.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190235.hg.1 |
NM_007146.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700190235.hg.1 |
ENST00000258963.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190235.hg.1 |
ENST00000581208.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190235.hg.1 |
ENST00000584396.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700190235.hg.1 |
NM_007146.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700190235.hg.1 |
TC1700011237.hg.1 |
Clariom D TC |