Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLE6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143986.1 | NP_001137458.1 | 8 - 9 | 669 | 63 | |
NM_024760.2 | NP_079036.1 | 7 - 8 | 591 | 63 | ||
XM_005259645.2 | XP_005259702.1 | 8 - 9 | 678 | 63 | ||
XM_011528300.2 | XP_011526602.1 | 8 - 9 | 654 | 63 | ||
XM_011528301.1 | XP_011526603.1 | 4 - 5 | 381 | 63 | ||
GenBank mRNA | AK024071.1 | - | 8 - 9 | 644 | 63 | |
BC007215.1 | - | 2 - 3 | 277 | 2824175 | 63 | |
BC013986.2 | - | 7 - 8 | 549 | 3503930 | 63 | |
BC020206.1 | - | 8 - 9 | 626 | 4621656 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |