Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003496.hg.1 |
ENST00000327813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003496.hg.1 |
ENST00000400335.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003496.hg.1 |
ENST00000400338.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003496.hg.1 |
ENST00000407188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003496.hg.1 |
NM_001039570.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003496.hg.1 |
NM_032045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003496.hg.1 |
TC22000191.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000327813.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000327813.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000327813.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000327813.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000327813.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000327813.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000327813.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000327813.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000327813.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000327813.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000327813.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000327813.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000327813.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000327813.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000327813.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000400335.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000400335.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000400335.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000400335.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000400335.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000400335.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000400335.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000400335.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000400335.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000400335.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000400335.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000400335.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000400335.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000400335.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000400335.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000407188.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000407188.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000407188.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000407188.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000407188.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000407188.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000407188.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000407188.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000407188.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000407188.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000407188.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000407188.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000407188.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000453585.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000453585.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000453585.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000453585.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000474001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000474001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000474001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000474001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000479755.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
ENST00000479755.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
NM_001039570.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
NM_001039570.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
NM_001039570.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
NM_001039570.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
NM_001039570.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
NM_001039570.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
NM_001039570.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
NM_001039570.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
NM_001039570.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
NM_001039570.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
NM_001039570.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
NM_001039570.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
NM_001039570.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
NM_001039570.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
NM_001039570.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
NM_032045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
NM_032045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
NM_032045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
NM_032045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
NM_032045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
NM_032045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
NM_032045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
NM_032045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
NM_032045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
NM_032045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
NM_032045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
NM_032045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
NM_032045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
NM_032045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
NM_032045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009248.hg.1 |
NM_032045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003494.hg.1 |
ENST00000327813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003494.hg.1 |
ENST00000400335.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003494.hg.1 |
ENST00000400338.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003494.hg.1 |
ENST00000407188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003494.hg.1 |
NM_001039570.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003494.hg.1 |
NM_032045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003494.hg.1 |
TC22000191.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000327813.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000327813.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000327813.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000327813.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000327813.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000327813.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000327813.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000327813.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000327813.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000327813.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000327813.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000327813.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000327813.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000327813.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000327813.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000400335.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000400335.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000400335.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000400335.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000400335.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000400335.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000400335.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000400335.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000400335.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000400335.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000400335.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000400335.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000400335.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000400335.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000400335.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000407188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000407188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000407188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000407188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000407188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000407188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000407188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000407188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000407188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000407188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000407188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000407188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000407188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000453585.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000453585.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000453585.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000453585.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000474001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000474001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000474001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000474001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000479755.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
ENST00000479755.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
NM_001039570.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
NM_001039570.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
NM_001039570.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
NM_001039570.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
NM_001039570.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
NM_001039570.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
NM_001039570.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
NM_001039570.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
NM_001039570.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
NM_001039570.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
NM_001039570.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
NM_001039570.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
NM_001039570.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
NM_001039570.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
NM_001039570.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
NM_032045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
NM_032045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
NM_032045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
NM_032045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
NM_032045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
NM_032045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
NM_032045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
NM_032045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
NM_032045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
NM_032045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
NM_032045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
NM_032045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
NM_032045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
NM_032045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
NM_032045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200009248.hg.1 |
NM_032045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200168394.hg.1 |
TC2200009248.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200168394.hg.1 |
ENST00000327813.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168394.hg.1 |
ENST00000400335.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168394.hg.1 |
ENST00000407188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168394.hg.1 |
ENST00000453585.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168394.hg.1 |
NM_001039570.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200168394.hg.1 |
NM_032045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200168394.hg.1 |
ENST00000327813.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168394.hg.1 |
ENST00000400335.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168394.hg.1 |
ENST00000407188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168394.hg.1 |
ENST00000453585.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168394.hg.1 |
NM_001039570.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200168394.hg.1 |
NM_032045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200168396.hg.1 |
TC2200009248.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200168396.hg.1 |
ENST00000327813.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168396.hg.1 |
ENST00000400335.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168396.hg.1 |
ENST00000407188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168396.hg.1 |
ENST00000453585.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168396.hg.1 |
ENST00000474001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168396.hg.1 |
NM_001039570.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200168396.hg.1 |
NM_032045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200168396.hg.1 |
ENST00000327813.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168396.hg.1 |
ENST00000400335.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168396.hg.1 |
ENST00000407188.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168396.hg.1 |
ENST00000453585.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168396.hg.1 |
ENST00000474001.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200168396.hg.1 |
NM_001039570.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200168396.hg.1 |
NM_032045.4 |
Refseq |