Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005345.hg.1 |
TC22001465.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005345.hg.1 |
ENST00000403663.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005345.hg.1 |
ENST00000406386.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005345.hg.1 |
ENST00000407319.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005345.hg.1 |
NM_001039141.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005345.hg.1 |
NM_007032.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22005345.hg.1 |
NM_138632.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200151391.hg.1 |
TC2200007313.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200151391.hg.1 |
ENST00000331103.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151391.hg.1 |
ENST00000344404.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151391.hg.1 |
ENST00000403663.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151391.hg.1 |
ENST00000406386.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151391.hg.1 |
ENST00000407319.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151391.hg.1 |
ENST00000413051.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151391.hg.1 |
ENST00000417857.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151391.hg.1 |
ENST00000418339.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151391.hg.1 |
ENST00000428075.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151391.hg.1 |
ENST00000452519.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151391.hg.1 |
ENST00000633257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151391.hg.1 |
NM_001039141.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200151391.hg.1 |
NM_007032.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200151391.hg.1 |
NM_138632.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200151391.hg.1 |
ENST00000331103.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151391.hg.1 |
ENST00000344404.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151391.hg.1 |
ENST00000403663.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151391.hg.1 |
ENST00000406386.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151391.hg.1 |
ENST00000407319.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151391.hg.1 |
ENST00000413051.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151391.hg.1 |
ENST00000417857.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151391.hg.1 |
ENST00000418339.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151391.hg.1 |
ENST00000428075.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151391.hg.1 |
ENST00000452519.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151391.hg.1 |
ENST00000633257.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200151391.hg.1 |
NM_001039141.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200151391.hg.1 |
NM_007032.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200151391.hg.1 |
NM_138632.2 |
Refseq |