Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003292.2 | NP_003283.2 | 35 - 36 | 5267 | 66 | |
GenBank mRNA | U69668.1 | - | 35 - 36 | 5058 | 66 | |
X66397.1 | - | 35 - 36 | 5267 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Detect Genomic DNA Homo sapiens Probe spans exons |