Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
ENST00000357650.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
ENST00000357650.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
ENST00000357650.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
ENST00000357650.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
ENST00000357650.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
ENST00000357650.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
ENST00000357650.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
ENST00000357650.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
ENST00000357650.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
ENST00000357650.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
ENST00000357650.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
ENST00000357650.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
ENST00000357650.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
ENST00000370111.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
ENST00000370111.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
ENST00000370111.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
ENST00000370112.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
ENST00000370112.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
ENST00000370112.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
ENST00000370112.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
ENST00000370112.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
ENST00000370112.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
ENST00000370112.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
ENST00000370112.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
ENST00000370112.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
ENST00000370112.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
ENST00000370112.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
ENST00000370112.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
ENST00000370112.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
ENST00000370112.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
ENST00000370112.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
ENST00000370112.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
NM_001144884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
NM_001144884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
NM_001144884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
NM_001144884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
NM_001144884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
NM_001144884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
NM_001144884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
NM_001144884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
NM_001144884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
NM_001144884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
NM_001144884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
NM_001144884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
NM_001144884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
NM_001144884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
NM_001144884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
NM_001144884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
NM_133496.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
NM_133496.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
NM_133496.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
NM_133496.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
NM_133496.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
NM_133496.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
NM_133496.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
NM_133496.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
NM_133496.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
NM_133496.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
NM_133496.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
NM_133496.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
NM_133496.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
NM_133496.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
NM_133496.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
NM_133496.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
NM_133496.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009226.hg.1 |
NM_133496.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014270.hg.1 |
TC01000905.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014270.hg.1 |
ENST00000357650.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014270.hg.1 |
ENST00000370112.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014270.hg.1 |
NM_001144884.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01014270.hg.1 |
NM_133496.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165719.hg.1 |
ENST00000357650.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165719.hg.1 |
ENST00000370112.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165719.hg.1 |
NM_001144884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165719.hg.1 |
NM_133496.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165719.hg.1 |
ENST00000357650.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165719.hg.1 |
ENST00000370112.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100165719.hg.1 |
NM_001144884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165719.hg.1 |
NM_133496.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100165719.hg.1 |
TC0100009226.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
ENST00000357650.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
ENST00000357650.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
ENST00000357650.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
ENST00000357650.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
ENST00000357650.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
ENST00000357650.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
ENST00000357650.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
ENST00000357650.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
ENST00000357650.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
ENST00000357650.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
ENST00000357650.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
ENST00000357650.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
ENST00000357650.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
ENST00000370111.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
ENST00000370111.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
ENST00000370111.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
ENST00000370112.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
ENST00000370112.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
ENST00000370112.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
ENST00000370112.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
ENST00000370112.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
ENST00000370112.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
ENST00000370112.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
ENST00000370112.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
ENST00000370112.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
ENST00000370112.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
ENST00000370112.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
ENST00000370112.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
ENST00000370112.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
ENST00000370112.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
ENST00000370112.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
ENST00000370112.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
NM_001144884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
NM_001144884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
NM_001144884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
NM_001144884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
NM_001144884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
NM_001144884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
NM_001144884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
NM_001144884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
NM_001144884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
NM_001144884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
NM_001144884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
NM_001144884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
NM_001144884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
NM_001144884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
NM_001144884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
NM_001144884.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
NM_133496.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
NM_133496.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
NM_133496.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
NM_133496.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
NM_133496.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
NM_133496.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
NM_133496.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
NM_133496.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
NM_133496.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
NM_133496.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
NM_133496.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
NM_133496.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
NM_133496.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
NM_133496.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
NM_133496.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
NM_133496.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
NM_133496.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100009226.hg.1 |
NM_133496.4 |
Refseq |