Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016463.hg.1 |
TC08001280.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016463.hg.1 |
ENST00000276569.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016463.hg.1 |
ENST00000458464.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08016463.hg.1 |
NM_018120.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000276569.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000276569.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000276569.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000276569.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000276569.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000276569.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000276569.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000276569.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000276569.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000276569.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000276569.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000276569.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000458464.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000458464.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000458464.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000458464.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000458464.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000458464.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000458464.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000458464.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000458464.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000458464.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000518908.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000518908.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000518908.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000518908.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000518908.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000518908.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000518908.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000518908.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000518908.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000519352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000519352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000519352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000519352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000519352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000519352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000519352.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000523384.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000523384.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000523384.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000523384.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000523384.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000528721.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000528721.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000528721.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000528721.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000528721.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000528721.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000528721.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
ENST00000528721.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
NM_001286702.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
NM_001286702.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
NM_001286702.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
NM_001286702.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
NM_001286702.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
NM_001286702.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
NM_001286702.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
NM_001286702.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
NM_001286702.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
NM_001286702.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
NM_018120.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
NM_018120.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
NM_018120.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
NM_018120.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
NM_018120.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
NM_018120.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
NM_018120.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
NM_018120.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
NM_018120.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
NM_018120.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
NM_018120.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800010663.hg.1 |
NM_018120.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172363.hg.1 |
ENST00000276569.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172363.hg.1 |
ENST00000458464.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172363.hg.1 |
ENST00000518908.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172363.hg.1 |
ENST00000519352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172363.hg.1 |
ENST00000523384.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172363.hg.1 |
ENST00000528721.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172363.hg.1 |
NM_001286702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172363.hg.1 |
NM_018120.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172363.hg.1 |
ENST00000276569.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172363.hg.1 |
ENST00000458464.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172363.hg.1 |
ENST00000518908.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172363.hg.1 |
ENST00000519352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172363.hg.1 |
ENST00000523384.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172363.hg.1 |
ENST00000528721.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172363.hg.1 |
NM_001286702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172363.hg.1 |
NM_018120.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172363.hg.1 |
TC0800010663.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800172362.hg.1 |
ENST00000276569.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172362.hg.1 |
ENST00000458464.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172362.hg.1 |
ENST00000518908.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172362.hg.1 |
ENST00000519352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172362.hg.1 |
ENST00000528721.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172362.hg.1 |
NM_001286702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172362.hg.1 |
NM_018120.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172362.hg.1 |
ENST00000276569.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172362.hg.1 |
ENST00000458464.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172362.hg.1 |
ENST00000518908.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172362.hg.1 |
ENST00000519352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172362.hg.1 |
ENST00000528721.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800172362.hg.1 |
NM_001286702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172362.hg.1 |
NM_018120.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800172362.hg.1 |
TC0800010663.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000276569.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000276569.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000276569.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000276569.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000276569.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000276569.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000276569.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000276569.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000276569.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000276569.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000276569.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000276569.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000458464.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000458464.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000458464.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000458464.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000458464.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000458464.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000458464.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000458464.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000458464.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000458464.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000518908.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000518908.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000518908.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000518908.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000518908.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000518908.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000518908.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000518908.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000518908.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000519352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000519352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000519352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000519352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000519352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000519352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000519352.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000523384.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000523384.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000523384.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000523384.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000523384.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000528721.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000528721.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000528721.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000528721.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000528721.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000528721.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000528721.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
ENST00000528721.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
NM_001286702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
NM_001286702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
NM_001286702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
NM_001286702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
NM_001286702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
NM_001286702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
NM_001286702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
NM_001286702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
NM_001286702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
NM_001286702.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
NM_018120.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
NM_018120.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
NM_018120.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
NM_018120.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
NM_018120.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
NM_018120.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
NM_018120.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
NM_018120.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
NM_018120.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
NM_018120.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
NM_018120.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800010663.hg.1 |
NM_018120.5 |
Refseq |