Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000309958.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000309958.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000309958.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000309958.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000309958.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000309958.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000309958.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000309958.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000309958.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000309958.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000309958.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000358789.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000358789.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000358789.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000358789.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000358789.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000358789.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000358789.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000358789.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000358789.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000358789.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000358789.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000391805.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000391805.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000391805.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000391805.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000391805.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000391805.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000391805.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000391805.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000391805.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000391805.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000391805.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000599077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000599077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000599077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000599077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000599987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000599987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000599987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000599987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000599987.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000601467.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000601467.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000601467.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000601467.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000601467.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000601467.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
ENST00000601467.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
NM_001077500.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
NM_001077500.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
NM_001077500.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
NM_001077500.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
NM_001077500.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
NM_001077500.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
NM_001077500.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
NM_001077500.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
NM_001077500.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
NM_001077500.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
NM_001077500.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
NM_002776.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
NM_002776.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
NM_002776.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
NM_002776.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
NM_002776.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
NM_002776.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
NM_002776.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
NM_002776.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
NM_002776.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
NM_002776.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
NM_002776.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
NM_145888.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
NM_145888.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
NM_145888.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
NM_145888.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
NM_145888.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
NM_145888.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
NM_145888.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
NM_145888.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
NM_145888.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
NM_145888.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011233.hg.1 |
NM_145888.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024027.hg.1 |
ENST00000309958.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024027.hg.1 |
NM_002776.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19024027.hg.1 |
TC19001767.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900194503.hg.1 |
ENST00000309958.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194503.hg.1 |
NM_002776.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900194503.hg.1 |
TC1900011233.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900194503.hg.1 |
ENST00000309958.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900194503.hg.1 |
NM_002776.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000309958.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000309958.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000309958.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000309958.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000309958.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000309958.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000309958.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000309958.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000309958.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000309958.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000309958.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000358789.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000358789.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000358789.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000358789.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000358789.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000358789.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000358789.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000358789.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000358789.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000358789.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000358789.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000391805.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000391805.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000391805.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000391805.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000391805.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000391805.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000391805.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000391805.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000391805.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000391805.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000391805.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000599077.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000599077.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000599077.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000599077.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000599987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000599987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000599987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000599987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000599987.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000601467.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000601467.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000601467.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000601467.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000601467.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000601467.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
ENST00000601467.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
NM_001077500.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
NM_001077500.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
NM_001077500.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
NM_001077500.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
NM_001077500.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
NM_001077500.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
NM_001077500.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
NM_001077500.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
NM_001077500.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
NM_001077500.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
NM_001077500.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
NM_002776.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
NM_002776.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
NM_002776.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
NM_002776.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
NM_002776.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
NM_002776.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
NM_002776.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
NM_002776.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
NM_002776.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
NM_002776.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
NM_002776.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
NM_145888.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
NM_145888.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
NM_145888.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
NM_145888.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
NM_145888.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
NM_145888.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
NM_145888.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
NM_145888.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
NM_145888.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
NM_145888.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011233.hg.1 |
NM_145888.2 |
Refseq |