Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027792.hg.1 |
ENST00000227665.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027792.hg.1 |
ENST00000542499.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027792.hg.1 |
NM_001166598.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027792.hg.1 |
NM_052968.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027792.hg.1 |
TC11002325.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027796.hg.1 |
ENST00000542499.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027796.hg.1 |
NM_001166598.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027796.hg.1 |
NM_052968.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027796.hg.1 |
TC11002325.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027791.hg.1 |
ENST00000227665.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027791.hg.1 |
ENST00000542499.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027791.hg.1 |
NM_001166598.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027791.hg.1 |
NM_052968.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11027791.hg.1 |
TC11002325.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012420.hg.1 |
ENST00000227665.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012420.hg.1 |
ENST00000227665.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012420.hg.1 |
ENST00000227665.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012420.hg.1 |
ENST00000227665.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012420.hg.1 |
ENST00000227665.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012420.hg.1 |
ENST00000227665.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012420.hg.1 |
ENST00000433069.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012420.hg.1 |
ENST00000433069.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012420.hg.1 |
ENST00000433069.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012420.hg.1 |
ENST00000433069.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012420.hg.1 |
ENST00000542499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012420.hg.1 |
ENST00000542499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012420.hg.1 |
ENST00000542499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012420.hg.1 |
ENST00000542499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012420.hg.1 |
ENST00000542499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012420.hg.1 |
ENST00000542499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012420.hg.1 |
ENST00000542499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012420.hg.1 |
NM_001166598.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012420.hg.1 |
NM_001166598.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012420.hg.1 |
NM_001166598.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012420.hg.1 |
NM_001166598.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012420.hg.1 |
NM_001166598.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012420.hg.1 |
NM_001166598.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012420.hg.1 |
NM_001166598.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012420.hg.1 |
NM_052968.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012420.hg.1 |
NM_052968.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012420.hg.1 |
NM_052968.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012420.hg.1 |
NM_052968.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012420.hg.1 |
NM_052968.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012420.hg.1 |
NM_052968.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012420.hg.1 |
NM_052968.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195369.hg.1 |
ENST00000227665.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195369.hg.1 |
ENST00000433069.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195369.hg.1 |
ENST00000542499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195369.hg.1 |
NM_001166598.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195369.hg.1 |
NM_052968.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195369.hg.1 |
ENST00000227665.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195369.hg.1 |
ENST00000433069.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195369.hg.1 |
ENST00000542499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195369.hg.1 |
NM_001166598.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195369.hg.1 |
NM_052968.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195369.hg.1 |
TC1100012420.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100012420.hg.1 |
ENST00000227665.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012420.hg.1 |
ENST00000227665.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012420.hg.1 |
ENST00000227665.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012420.hg.1 |
ENST00000227665.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012420.hg.1 |
ENST00000227665.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012420.hg.1 |
ENST00000227665.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012420.hg.1 |
ENST00000433069.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012420.hg.1 |
ENST00000433069.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012420.hg.1 |
ENST00000433069.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012420.hg.1 |
ENST00000433069.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012420.hg.1 |
ENST00000542499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012420.hg.1 |
ENST00000542499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012420.hg.1 |
ENST00000542499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012420.hg.1 |
ENST00000542499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012420.hg.1 |
ENST00000542499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012420.hg.1 |
ENST00000542499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012420.hg.1 |
ENST00000542499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012420.hg.1 |
NM_001166598.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012420.hg.1 |
NM_001166598.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012420.hg.1 |
NM_001166598.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012420.hg.1 |
NM_001166598.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012420.hg.1 |
NM_001166598.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012420.hg.1 |
NM_001166598.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012420.hg.1 |
NM_001166598.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012420.hg.1 |
NM_052968.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012420.hg.1 |
NM_052968.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012420.hg.1 |
NM_052968.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012420.hg.1 |
NM_052968.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012420.hg.1 |
NM_052968.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012420.hg.1 |
NM_052968.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012420.hg.1 |
NM_052968.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195370.hg.1 |
ENST00000227665.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195370.hg.1 |
ENST00000433069.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195370.hg.1 |
ENST00000542499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195370.hg.1 |
NM_001166598.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195370.hg.1 |
NM_052968.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195370.hg.1 |
ENST00000227665.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195370.hg.1 |
ENST00000433069.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195370.hg.1 |
ENST00000542499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195370.hg.1 |
NM_001166598.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195370.hg.1 |
NM_052968.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195370.hg.1 |
TC1100012420.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100195375.hg.1 |
ENST00000433069.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195375.hg.1 |
ENST00000542499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195375.hg.1 |
NM_001166598.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195375.hg.1 |
NM_052968.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195375.hg.1 |
ENST00000433069.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195375.hg.1 |
ENST00000542499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195375.hg.1 |
NM_001166598.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195375.hg.1 |
NM_052968.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195375.hg.1 |
TC1100012420.hg.1 |
Clariom D TC |