Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017546.hg.1 |
ENST00000343782.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017546.hg.1 |
ENST00000355254.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017546.hg.1 |
ENST00000357724.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017546.hg.1 |
ENST00000360948.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017546.hg.1 |
ENST00000394480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017546.hg.1 |
ENST00000542733.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017546.hg.1 |
ENST00000557856.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017546.hg.1 |
ENST00000558676.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017546.hg.1 |
NM_001012338.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017546.hg.1 |
NM_001243101.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017546.hg.1 |
NM_002530.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017546.hg.1 |
TC15001815.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017543.hg.1 |
ENST00000355254.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017543.hg.1 |
ENST00000357724.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017543.hg.1 |
ENST00000360948.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017543.hg.1 |
ENST00000394480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017543.hg.1 |
ENST00000542733.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017543.hg.1 |
ENST00000557856.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017543.hg.1 |
ENST00000558576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017543.hg.1 |
ENST00000558676.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017543.hg.1 |
NM_001012338.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017543.hg.1 |
NM_001243101.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017543.hg.1 |
NM_002530.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017543.hg.1 |
TC15001815.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017545.hg.1 |
ENST00000343782.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017545.hg.1 |
ENST00000355254.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017545.hg.1 |
ENST00000357724.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017545.hg.1 |
ENST00000360948.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017545.hg.1 |
ENST00000394480.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017545.hg.1 |
ENST00000542733.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017545.hg.1 |
ENST00000557856.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017545.hg.1 |
ENST00000558576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017545.hg.1 |
ENST00000558676.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017545.hg.1 |
NM_001012338.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017545.hg.1 |
NM_001243101.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017545.hg.1 |
NM_002530.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15017545.hg.1 |
TC15001815.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1500177866.hg.1 |
ENST00000355254.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177866.hg.1 |
ENST00000357724.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177866.hg.1 |
ENST00000360948.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177866.hg.1 |
ENST00000394480.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177866.hg.1 |
ENST00000542733.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177866.hg.1 |
ENST00000557856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177866.hg.1 |
ENST00000558676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177866.hg.1 |
ENST00000559680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177866.hg.1 |
ENST00000626019.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177866.hg.1 |
ENST00000629765.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177866.hg.1 |
NM_001012338.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500177866.hg.1 |
NM_001243101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500177866.hg.1 |
NM_002530.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500177866.hg.1 |
ENST00000355254.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177866.hg.1 |
ENST00000357724.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177866.hg.1 |
ENST00000360948.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177866.hg.1 |
ENST00000394480.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177866.hg.1 |
ENST00000542733.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177866.hg.1 |
ENST00000557856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177866.hg.1 |
ENST00000558676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177866.hg.1 |
ENST00000559680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177866.hg.1 |
ENST00000626019.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177866.hg.1 |
ENST00000629765.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177866.hg.1 |
NM_001012338.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500177866.hg.1 |
NM_001243101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500177866.hg.1 |
NM_002530.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500177866.hg.1 |
TC1500010350.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500177863.hg.1 |
ENST00000355254.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177863.hg.1 |
ENST00000357724.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177863.hg.1 |
ENST00000360948.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177863.hg.1 |
ENST00000394480.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177863.hg.1 |
ENST00000542733.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177863.hg.1 |
ENST00000557856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177863.hg.1 |
ENST00000558576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177863.hg.1 |
ENST00000558676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177863.hg.1 |
ENST00000626019.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177863.hg.1 |
ENST00000629765.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177863.hg.1 |
NM_001012338.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500177863.hg.1 |
NM_001243101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500177863.hg.1 |
NM_002530.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500177863.hg.1 |
ENST00000355254.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177863.hg.1 |
ENST00000357724.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177863.hg.1 |
ENST00000360948.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177863.hg.1 |
ENST00000394480.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177863.hg.1 |
ENST00000542733.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177863.hg.1 |
ENST00000557856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177863.hg.1 |
ENST00000558576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177863.hg.1 |
ENST00000558676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177863.hg.1 |
ENST00000626019.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177863.hg.1 |
ENST00000629765.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177863.hg.1 |
NM_001012338.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500177863.hg.1 |
NM_001243101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500177863.hg.1 |
NM_002530.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500177863.hg.1 |
TC1500010350.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500177865.hg.1 |
ENST00000355254.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177865.hg.1 |
ENST00000357724.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177865.hg.1 |
ENST00000360948.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177865.hg.1 |
ENST00000394480.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177865.hg.1 |
ENST00000542733.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177865.hg.1 |
ENST00000557856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177865.hg.1 |
ENST00000558576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177865.hg.1 |
ENST00000558676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177865.hg.1 |
ENST00000559680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177865.hg.1 |
ENST00000626019.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177865.hg.1 |
ENST00000629765.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177865.hg.1 |
NM_001012338.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500177865.hg.1 |
NM_001243101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500177865.hg.1 |
NM_002530.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500177865.hg.1 |
ENST00000355254.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177865.hg.1 |
ENST00000357724.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177865.hg.1 |
ENST00000360948.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177865.hg.1 |
ENST00000394480.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177865.hg.1 |
ENST00000542733.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177865.hg.1 |
ENST00000557856.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177865.hg.1 |
ENST00000558576.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177865.hg.1 |
ENST00000558676.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177865.hg.1 |
ENST00000559680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177865.hg.1 |
ENST00000626019.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177865.hg.1 |
ENST00000629765.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500177865.hg.1 |
NM_001012338.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500177865.hg.1 |
NM_001243101.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500177865.hg.1 |
NM_002530.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500177865.hg.1 |
TC1500010350.hg.1 |
Clariom D TC |